missing translation for 'onlineSavingsMsg'
Learn More

4-(Cyclopropylamino)benzonitrile, 98%, Thermo Scientific™

Product Code. 15477798
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1019607-55-6
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.204
InChI Key SHZXUUVOVQTNCK-UHFFFAOYSA-N
Synonym 4-cyclopropylamino benzonitrile, acmc-2097xs
PubChem CID 28411077
IUPAC Name 4-(cyclopropylamino)benzonitrile
SMILES C1CC1NC2=CC=C(C=C2)C#N

Specifications

Quantity 1 g
UN Number UN3439
Formula Weight 158.2
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-(Cyclopropylamino)benzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'