missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanopyridine, 98%, Thermo Scientific Chemicals

£21.51 - £288.44

Chemical Identifiers

CAS 100-48-1
Molecular Formula C6H4N2
Molecular Weight (g/mol) 104.112
MDL Number MFCD00006417
InChI Key GPHQHTOMRSGBNZ-UHFFFAOYSA-N
Synonym 4-cyanopyridine, isonicotinonitrile, 4-pyridinecarbonitrile, isonicotinic acid nitrile, 4-pyridinenitrile, 4-cyano pyridine, .gamma.-cyanopyridine, gamma-cyanopyridine, p-cyanopyridine, pubchem11155
PubChem CID 7506
ChEBI CHEBI:28020
IUPAC Name pyridine-4-carbonitrile
SMILES C1=CN=CC=C1C#N
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11364739
SDS View Documents
Thermo Scientific Alfa Aesar
A10286.09
10 g
£21.51
10g
Please to purchase this item. Need a web account? Register with us today!
11414023
SDS View Documents
Thermo Scientific Alfa Aesar
A10286.22
100 g
£25.43
100g
Please to purchase this item. Need a web account? Register with us today!
11424023
SDS View Documents
Thermo Scientific Alfa Aesar
A10286.36
500 g
£77.08
500g
Please to purchase this item. Need a web account? Register with us today!
11434023
SDS View Documents
Thermo Scientific Alfa Aesar
A10286.0E
2500 g
£288.44
2500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

4-Cyanopyridine is used as an intermediate in organic synthesis and pharmaceutical substances like isonicotinylhydrazide, which is used in the treatment of tuberculosis. It is used as a precursor for the preparation of isonicotinic acid and 4-diemthylaminopyridine (DMAP). It is involved in the synthesis of 6-methyl-2-pyridin-4-yl-pyrimidin-4-ylamine by reacting with acetonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyanopyridine is used as an intermediate in organic synthesis and pharmaceutical substances like isonicotinylhydrazide, which is used in the treatment of tuberculosis. It is used as a precursor for the preparation of isonicotinic acid and 4-diemthylaminopyridine (DMAP). It is involved in the synthesis of 6-methyl-2-pyridin-4-yl-pyrimidin-4-ylamine by reacting with acetonitrile.

Solubility
Soluble in water.

Notes
Incompatible with strong acids, strong bases and strong oxidizing agents.
Specifications

Chemical Identifiers

100-48-1
104.112
GPHQHTOMRSGBNZ-UHFFFAOYSA-N
7506
pyridine-4-carbonitrile
C6H4N2
MFCD00006417
4-cyanopyridine, isonicotinonitrile, 4-pyridinecarbonitrile, isonicotinic acid nitrile, 4-pyridinenitrile, 4-cyano pyridine, .gamma.-cyanopyridine, gamma-cyanopyridine, p-cyanopyridine, pubchem11155
CHEBI:28020
C1=CN=CC=C1C#N

Specifications

100-48-1
6
88°C (190°F)
C6H4N2
107712
Soluble in water.
C1=CN=CC=C1C#N
104.112
CHEBI:28020
98%
76°C to 80°C
194°C to 196°C
Pungent
MFCD00006417
4-cyanopyridine, isonicotinonitrile, 4-pyridinecarbonitrile, isonicotinic acid nitrile, 4-pyridinenitrile, 4-cyano pyridine, .gamma.-cyanopyridine, gamma-cyanopyridine, p-cyanopyridine, pubchem11155
GPHQHTOMRSGBNZ-UHFFFAOYSA-N
pyridine-4-carbonitrile
7506
104.11
4-Cyanopyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.

P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H302-H312-H315-H319-H332-H335

missing translation for 'einecsNumber' : 202-856-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.