missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanopiperidine, 97%, Thermo Scientific Chemicals

£45.33 - £143.72

Chemical Identifiers

CAS 4395-98-6
Molecular Formula C6H10N2
Molecular Weight (g/mol) 110.16
InChI Key FSDNTQSJGHSJBG-UHFFFAOYSA-N
Synonym 4-cyanopiperidine, 4-piperidinecarbonitrile, 4-cyanopiperdine, 4-cyanopyperidine, 4-cyano piperdine, 4-cyano piperidine, 4-cyano-piperidine, acmc-20aipl, pubchem9349
PubChem CID 138223
IUPAC Name piperidine-4-carbonitrile
SMILES C1CNCCC1C#N
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
11966411
SDS View Documents
Thermo Scientific Acros
441820050
5 g Glass bottle
£45.33
5g
Please to purchase this item. Need a web account? Register with us today!
11986411
SDS View Documents
Thermo Scientific Acros
441820250
25 g Glass bottle
£143.72
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4395-98-6
110.16
4-cyanopiperidine, 4-piperidinecarbonitrile, 4-cyanopiperdine, 4-cyanopyperidine, 4-cyano piperdine, 4-cyano piperidine, 4-cyano-piperidine, acmc-20aipl, pubchem9349
piperidine-4-carbonitrile
C6H10N2
FSDNTQSJGHSJBG-UHFFFAOYSA-N
138223
C1CNCCC1C#N

Specifications

4395-98-6
100.0
100.0°C to 105.0°C (12.0 mmHg)
96% min. (GC)
4-cyanopiperidine, 4-piperidinecarbonitrile, 4-cyanopiperdine, 4-cyanopyperidine, 4-cyano piperdine, 4-cyano piperidine, 4-cyano-piperidine, acmc-20aipl, pubchem9349
C1CNCCC1C#N
110.16
110.16
Liquid
96.0
Colorless to Yellow
Authentic
C6H10N2
FSDNTQSJGHSJBG-UHFFFAOYSA-N
piperidine-4-carbonitrile
138223
97%
4-Cyanopiperidine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Danger

Product Identifier
  • 4-Cyanopiperidine, 97%

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.