missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-3-iodopyridine, 98%, Thermo Scientific Chemicals

£109.00 - £268.00

Chemical Identifiers

CAS 10386-28-4
Molecular Formula C6H3IN2
Molecular Weight (g/mol) 230.008
MDL Number MFCD08729285
InChI Key KMFIQUYSWTUCGD-UHFFFAOYSA-N
Synonym 3-iodoisonicotinonitrile, 4-cyano-3-iodopyridine, 3-iodo-4-pyridinecarbonitrile, 3-iodo-isonicotinonitrile, acmc-1c8uo, 4-pyridinecarbonitrile,3-iodo, 3-iodanylpyridine-4-carbonitrile
PubChem CID 12180887
IUPAC Name 3-iodopyridine-4-carbonitrile
SMILES C1=CN=CC(=C1C#N)I
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15413647
View Documents
Thermo Scientific Alfa Aesar
H27247.MD
250 mg
£109.00
250mg
Please to purchase this item. Need a web account? Register with us today!
11322269
View Documents
Thermo Scientific Alfa Aesar
H27247.03
1 g
£268.00
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10386-28-4
230.008
KMFIQUYSWTUCGD-UHFFFAOYSA-N
12180887
C1=CN=CC(=C1C#N)I
C6H3IN2
MFCD08729285
3-iodoisonicotinonitrile, 4-cyano-3-iodopyridine, 3-iodo-4-pyridinecarbonitrile, 3-iodo-isonicotinonitrile, acmc-1c8uo, 4-pyridinecarbonitrile,3-iodo, 3-iodanylpyridine-4-carbonitrile
3-iodopyridine-4-carbonitrile

Specifications

10386-28-4
C6H3IN2
UN3439
3-iodoisonicotinonitrile, 4-cyano-3-iodopyridine, 3-iodo-4-pyridinecarbonitrile, 3-iodo-isonicotinonitrile, acmc-1c8uo, 4-pyridinecarbonitrile,3-iodo, 3-iodanylpyridine-4-carbonitrile
C1=CN=CC(=C1C#N)I
230.008
230.01
4-Cyano-3-iodopyridine
119°C to 122°C
MFCD08729285
Light sensitive
KMFIQUYSWTUCGD-UHFFFAOYSA-N
3-iodopyridine-4-carbonitrile
12180887
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only