missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorobenzene-1-carboximidamide hydrochloride, 95+%, Thermo Scientific™
£23.00 - £222.00
Chemical Identifiers
CAS | 14401-51-5 |
---|---|
Molecular Formula | C7H8Cl2N2 |
Molecular Weight (g/mol) | 191.055 |
InChI Key | RXAOGVQDNBYURA-UHFFFAOYSA-N |
Synonym | 4-chlorobenzamidine hydrochloride, 4-chlorobenzene-1-carboximidamide hydrochloride, 4-chlorobenzamidine hcl, p-chlorobenzamidine hydrochloride, 4-chloro-benzamidine hydrochloride, 4-chlorobenzenecarboximidamide monohydrochloride, benzenecarboximidamide, 4-chloro-, monohydrochloride, 4-chlorobenzimidamide hydrochloride, benzamide, p-chloro-, hydrochloride 7ci, benzamidine, p-chloro-, monohydrochloride |
PubChem CID | 203436 |
IUPAC Name | 4-chlorobenzenecarboximidamide;hydrochloride |
SMILES | C1=CC(=CC=C1C(=N)N)Cl.Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10263622
|
Thermo Scientific Maybridge
SPB06264DA |
1 g | Amber Glass Bottle |
£23.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10673233
|
Thermo Scientific Maybridge
SPB06264EA |
10 g | Amber Glass Bottle |
£170.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10223203
|
Thermo Scientific Maybridge
SPB06264EB |
25 g | Amber Glass Bottle |
£222.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
14401-51-5 | |
191.055 | |
4-chlorobenzamidine hydrochloride, 4-chlorobenzene-1-carboximidamide hydrochloride, 4-chlorobenzamidine hcl, p-chlorobenzamidine hydrochloride, 4-chloro-benzamidine hydrochloride, 4-chlorobenzenecarboximidamide monohydrochloride, benzenecarboximidamide, 4-chloro-, monohydrochloride, 4-chlorobenzimidamide hydrochloride, benzamide, p-chloro-, hydrochloride 7ci, benzamidine, p-chloro-, monohydrochloride | |
4-chlorobenzenecarboximidamide;hydrochloride |
C7H8Cl2N2 | |
RXAOGVQDNBYURA-UHFFFAOYSA-N | |
203436 | |
C1=CC(=CC=C1C(=N)N)Cl.Cl |
Specifications
14401-51-5 | |
C7H8Cl2N2 | |
4-chlorobenzamidine hydrochloride, 4-chlorobenzene-1-carboximidamide hydrochloride, 4-chlorobenzamidine hcl, p-chlorobenzamidine hydrochloride, 4-chloro-benzamidine hydrochloride, 4-chlorobenzenecarboximidamide monohydrochloride, benzenecarboximidamide, 4-chloro-, monohydrochloride, 4-chlorobenzimidamide hydrochloride, benzamide, p-chloro-, hydrochloride 7ci, benzamidine, p-chloro-, monohydrochloride | |
C1=CC(=CC=C1C(=N)N)Cl.Cl | |
191.055 | |
191.06 | |
4-Chlorobenzene-1-carboximidamide hydrochloride |
95% | |
ClC6H4C(=NH)NH2.HCl | |
RXAOGVQDNBYURA-UHFFFAOYSA-N | |
4-chlorobenzenecarboximidamide;hydrochloride | |
203436 | |
95% |
Safety and Handling
Warning