missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2,6-difluorophenol, 97%, Thermo Scientific™

Product Code. 11461270
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 164790-68-5
Molecular Formula C6H3ClF2O
Molecular Weight (g/mol) 164.54
MDL Number MFCD06660242
InChI Key BVXYFECYLILSJV-UHFFFAOYSA-N
Synonym phenol, 4-chloro-2,6-difluoro, 2,6-difluoro-4-chlorophenol, pubchem22725, acmc-209drt, intermediates-zcf02236
PubChem CID 17750691
IUPAC Name 4-chloro-2,6-difluorophenol
SMILES OC1=C(F)C=C(Cl)C=C1F

Specifications

Quantity 5 g
UN Number UN3261
Formula Weight 164.54
Percent Purity 97%
Chemical Name or Material 4-Chloro-2,6-difluorophenol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'