Learn More
4-Carboxyphenylboronic acid, 97%, Thermo Scientific Chemicals
£24.92 - £206.03
Chemical Identifiers
CAS | 14047-29-1 |
---|---|
Molecular Formula | C7H7BO4 |
Molecular Weight (g/mol) | 165.94 |
MDL Number | MFCD00151801 |
InChI Key | SIAVMDKGVRXFAX-UHFFFAOYSA-N |
Synonym | 4-carboxyphenylboronic acid, 4-carboxybenzeneboronic acid, 4-dihydroxyboranyl benzoic acid, benzoic acid, 4-borono, 4-dihydroxyboryl benzoic acid, p-carboxyphenylboronic acid, 4-phenylester boronic acid, 4-carboxyphenyl boronic acid, chembl82324 |
PubChem CID | 312183 |
IUPAC Name | 4-boronobenzoic acid |
SMILES | OB(O)C1=CC=C(C=C1)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10556261
|
Thermo Scientific Acros
344400010 |
1 g | Glass bottle |
£24.92
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10566261
|
Thermo Scientific Acros
344400050 |
5 g | Glass bottle |
£62.51
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10626194
|
Thermo Scientific Acros
344400250 |
25 g | Glass bottle |
£206.03
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14047-29-1 | |
165.94 | |
SIAVMDKGVRXFAX-UHFFFAOYSA-N | |
312183 | |
OB(O)C1=CC=C(C=C1)C(O)=O |
C7H7BO4 | |
MFCD00151801 | |
4-carboxyphenylboronic acid, 4-carboxybenzeneboronic acid, 4-dihydroxyboranyl benzoic acid, benzoic acid, 4-borono, 4-dihydroxyboryl benzoic acid, p-carboxyphenylboronic acid, 4-phenylester boronic acid, 4-carboxyphenyl boronic acid, chembl82324 | |
4-boronobenzoic acid |
Specifications
14047-29-1 | |
100.0 | |
White to Beige | |
97% | |
HOOCC6H4B(OH)2 | |
4-carboxyphenylboronic acid, 4-carboxybenzeneboronic acid, 4-dihydroxyboranyl benzoic acid, benzoic acid, 4-borono, 4-dihydroxyboryl benzoic acid, p-carboxyphenylboronic acid, 4-phenylester boronic acid, 4-carboxyphenyl boronic acid, chembl82324 | |
SIAVMDKGVRXFAX-UHFFFAOYSA-N | |
4-boronobenzoic acid | |
312183 | |
97% | |
4-Carboxyphenylboronic acid |
96.0 | |
238.0°C to 240.0°C | |
Authentic | |
C7H7BO4 | |
MFCD00151801 | |
Solubility in water: 25g/L. | |
OB(O)C1=CC=C(C=C1)C(O)=O | |
165.94 | |
165.94 | |
Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and
GHS Signal Word: Warning
- 4-Carboxyphenylboronic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only