missing translation for 'onlineSavingsMsg'
Learn More

4'-Benzyloxyacetophenone, 98%, Thermo Scientific Chemicals

Product Code. 11493847
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54696-05-8
Molecular Formula C15H14O2
Molecular Weight (g/mol) 226.275
MDL Number MFCD00017247
InChI Key MKYMYZJJFMPDOA-UHFFFAOYSA-N
Synonym 4'-benzyloxyacetophenone, 1-4-benzyloxy phenyl ethanone, 4-benzyloxyacetophenone, 4'-benzyloxy-acetophenone, 1-4-benzyloxy phenyl ethan-1-one, ethanone, 1-4-phenylmethoxy phenyl, 4'-benzyloxy acetophenone, 1-4-benzyloxyphenyl ethanone
PubChem CID 245226
IUPAC Name 1-(4-phenylmethoxyphenyl)ethanone
SMILES CC(=O)C1=CC=C(C=C1)OCC2=CC=CC=C2

Specifications

Melting Point 91°C to 92°C
Quantity 100 g
Beilstein 1912124
Formula Weight 226.28
Percent Purity 98%
Chemical Name or Material 4'-Benzyloxyacetophenone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.