missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Methylphenoxy)piperidine, 99%, Thermo Scientific™

Catalog Number p-7033795
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
63843-49-2
C12H18ClNO
227.732
MFCD04115017
FUYMHJIAYHLIEH-UHFFFAOYSA-N
hydron; 4-4-methylphenoxy piperidine; chloride, hydrogen 4-4-methylphenoxy piperidine chloride
51066499
hydron;4-(4-methylphenoxy)piperidine;chloride
[H+].CC1=CC=C(C=C1)OC2CCNCC2.[Cl-]
This item is not returnable. View return policy
63843-49-2
C12H18ClNO
227.732
MFCD04115017
FUYMHJIAYHLIEH-UHFFFAOYSA-N
hydron; 4-4-methylphenoxy piperidine; chloride, hydrogen 4-4-methylphenoxy piperidine chloride
51066499
hydron;4-(4-methylphenoxy)piperidine;chloride
[H+].CC1=CC=C(C=C1)OC2CCNCC2.[Cl-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 191.28
Percent Purity 99%
Chemical Name or Material 4-(4-Methylphenoxy)piperidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'