missing translation for 'onlineSavingsMsg'
Learn More

4,4-Dimethylcyclohexanone, 97%, Thermo Scientific Chemicals

Product Code. 13488628
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4255-62-3
InChI Key PXQMSTLNSHMSJB-UHFFFAOYSA-N
Synonym 4,4-dimethylcyclohexanone, cyclohexanone, 4,4-dimethyl, 4,4-dimethyl cyclohexanone, 4,4-dimethyl-cyclohexan-1-one, 4,4-dimethycyclohexanone, 4.4-dimethylcyclohexanone, acmc-209jq8, 4,4-dimethyl-cyclohexanone, ksc494a0f, 4,4-dimethyl-1-cyclohexanone
PubChem CID 138166
IUPAC Name 4,4-dimethylcyclohexan-1-one
SMILES CC1(CCC(=O)CC1)C

Specifications

Melting Point 41°C to 43°C
Boiling Point 73°C (14.0 mmHg)
Flash Point 62°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C8H14O
Linear Formula (CH3)2C6H8(=O)
Quantity 250 mg
Solubility Information Solubility in water: insoluble
Molecular Weight (g/mol) 126.2
Formula Weight 126.2
Percent Purity 97%
Chemical Name or Material 4, 4-Dimethylcyclohexanone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4,4-Dimethylcyclohexanone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 1
Hazard Statement
  • H319-Causes serious eye irritation.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.