missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-[(3-Oxo-2,3-dihydro-4H-1,4-benzoxazin-4-yl)methyl]benzoic acid, Alfa Aesar™
Click to view available options
Quantity:
250 mg
1 g
Specifications
Quantity | 250 mg |
Formula Weight | 283.29 |
Chemical Name or Material | 4-[(3-Oxo-2,3-dihydro-4H-1,4-benzoxazin-4-yl)methyl]benzoic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'