Learn More
4-(3-Fluorophenylcarbamoyl)benzeneboronic acid, 98%, Thermo Scientific™
POA - £23.31
Chemical Identifiers
CAS | 874288-05-8 |
---|---|
Molecular Formula | C13H11BFNO3 |
Molecular Weight (g/mol) | 259.04 |
MDL Number | MFCD09027204 |
InChI Key | CVYDMRNYGKMSDI-UHFFFAOYSA-N |
Synonym | n-3-fluorophenyl 4-boronobenzamide, 4-3-fluorophenyl carbamoyl phenyl boronic acid, 4-borono-n-3-fluorophenyl benzamide, 4-3-fluorophenylcarbamoyl phenylboronic acid, 4-3-fluorophenyl carbamoyl phenylboronic acid, 4-3-fluorophenyl carbamoyl benzeneboronic acid, acmc-209qlr, n3-fluorophenyl 4-boronobenzamide |
PubChem CID | 44717609 |
IUPAC Name | [4-[(3-fluorophenyl)carbamoyl]phenyl]boronic acid |
SMILES | OB(O)C1=CC=C(C=C1)C(=O)NC1=CC(F)=CC=C1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
874288-05-8 | |
259.04 | |
CVYDMRNYGKMSDI-UHFFFAOYSA-N | |
44717609 | |
OB(O)C1=CC=C(C=C1)C(=O)NC1=CC(F)=CC=C1 |
C13H11BFNO3 | |
MFCD09027204 | |
n-3-fluorophenyl 4-boronobenzamide, 4-3-fluorophenyl carbamoyl phenyl boronic acid, 4-borono-n-3-fluorophenyl benzamide, 4-3-fluorophenylcarbamoyl phenylboronic acid, 4-3-fluorophenyl carbamoyl phenylboronic acid, 4-3-fluorophenyl carbamoyl benzeneboronic acid, acmc-209qlr, n3-fluorophenyl 4-boronobenzamide | |
[4-[(3-fluorophenyl)carbamoyl]phenyl]boronic acid |
Specifications
874288-05-8 | |
MFCD09027204 | |
CVYDMRNYGKMSDI-UHFFFAOYSA-N | |
[4-[(3-fluorophenyl)carbamoyl]phenyl]boronic acid | |
44717609 | |
98% |
C13H11BFNO3 | |
n-3-fluorophenyl 4-boronobenzamide, 4-3-fluorophenyl carbamoyl phenyl boronic acid, 4-borono-n-3-fluorophenyl benzamide, 4-3-fluorophenylcarbamoyl phenylboronic acid, 4-3-fluorophenyl carbamoyl phenylboronic acid, 4-3-fluorophenyl carbamoyl benzeneboronic acid, acmc-209qlr, n3-fluorophenyl 4-boronobenzamide | |
OB(O)C1=CC=C(C=C1)C(=O)NC1=CC(F)=CC=C1 | |
259.04 | |
259.04 | |
4-(3-Fluorophenylcarbamoyl)benzeneboronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.