Learn More
4-(2-Thiazolyl)phenol, 97%, Thermo Scientific Chemicals
£37.00 - £215.00
Chemical Identifiers
CAS | 81015-49-8 |
---|---|
Molecular Formula | C9H7NOS |
Molecular Weight (g/mol) | 177.221 |
MDL Number | MFCD07368615 |
InChI Key | LMMPCMRUGFBCPV-UHFFFAOYSA-N |
Synonym | 4-2-thiazolyl phenol, 4-1,3-thiazol-2-yl phenol, 4-thiazol-2-yl phenol, thiazole,2-4-trifluoromethyl phenyl, 4-3h-1,3-thiazol-2-ylidene cyclohexa-2,5-dien-1-one, 4-thiazol-2-yl-phenol, 4-thiazole-2-yl phenol, acmc-20a0c2, 4-thiazol-2-vl-phenol |
PubChem CID | 5354497 |
IUPAC Name | 4-(3H-1,3-thiazol-2-ylidene)cyclohexa-2,5-dien-1-one |
SMILES | C1=CC(=O)C=CC1=C2NC=CS2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15494467
|
Thermo Scientific Alfa Aesar
H31827.03 |
1 g |
£37.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15404477
|
Thermo Scientific Alfa Aesar
H31827.09 |
10 g |
£215.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
81015-49-8 | |
177.221 | |
LMMPCMRUGFBCPV-UHFFFAOYSA-N | |
5354497 | |
C1=CC(=O)C=CC1=C2NC=CS2 |
C9H7NOS | |
MFCD07368615 | |
4-2-thiazolyl phenol, 4-1,3-thiazol-2-yl phenol, 4-thiazol-2-yl phenol, thiazole,2-4-trifluoromethyl phenyl, 4-3h-1,3-thiazol-2-ylidene cyclohexa-2,5-dien-1-one, 4-thiazol-2-yl-phenol, 4-thiazole-2-yl phenol, acmc-20a0c2, 4-thiazol-2-vl-phenol | |
4-(3H-1,3-thiazol-2-ylidene)cyclohexa-2,5-dien-1-one |
Specifications
81015-49-8 | |
C9H7NOS | |
4-2-thiazolyl phenol, 4-1,3-thiazol-2-yl phenol, 4-thiazol-2-yl phenol, thiazole,2-4-trifluoromethyl phenyl, 4-3h-1,3-thiazol-2-ylidene cyclohexa-2,5-dien-1-one, 4-thiazol-2-yl-phenol, 4-thiazole-2-yl phenol, acmc-20a0c2, 4-thiazol-2-vl-phenol | |
C1=CC(=O)C=CC1=C2NC=CS2 | |
177.221 | |
177.28 | |
4-(2-Thiazolyl)phenol |
162°C to 165°C | |
MFCD07368615 | |
LMMPCMRUGFBCPV-UHFFFAOYSA-N | |
4-(3H-1,3-thiazol-2-ylidene)cyclohexa-2,5-dien-1-one | |
5354497 | |
97% |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.