missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Methylphenoxy)piperidine, 97%, Thermo Scientific Chemicals

Product Code. 11419700
Click to view available options
Quantity:
250 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63843-42-5
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.274
MDL Number MFCD06248344
InChI Key MLLZZLRYRWFFRQ-UHFFFAOYSA-N
Synonym 4-o-tolyloxy piperidine, 4-2-methylphenoxy piperidine, 4-o-tolyloxy-piperidine, acmc-1b7xi, piperidine,4-2-methylphenoxy
PubChem CID 3686170
IUPAC Name 4-(2-methylphenoxy)piperidine
SMILES CC1=CC=CC=C1OC2CCNCC2

Specifications

Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 191.28
Percent Purity 97%
Chemical Name or Material 4-(2-Methylphenoxy)piperidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'