missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(1H-Pyrazol-1-yl)benzaldehyde, ≥95%, Thermo Scientific™
Chemical Identifiers
CAS | 99662-34-7 |
---|---|
Molecular Formula | C10H8N2O |
Molecular Weight (g/mol) | 172.187 |
MDL Number | MFCD02681938 |
InChI Key | PPGRDLZPSDHBIC-UHFFFAOYSA-N |
Synonym | 4-1h-pyrazol-1-yl benzaldehyde, 4-pyrazol-1-yl-benzaldehyde, 1-4'-formylphenyl pyrazole, 1-4-formylphenyl pyrazole, 4-1h-pyrazol-1-yl benzenecarbaldehyde, 4-pyrazol-1-yl benzaldehyde, 1-4-formylphenyl-1h-pyrazole, benzaldehyde, 4-1h-pyrazol-1-yl, 4-pyrazolylbenzaldehyde, pubchem22713 |
PubChem CID | 2776477 |
IUPAC Name | 4-pyrazol-1-ylbenzaldehyde |
SMILES | C1=CN(N=C1)C2=CC=C(C=C2)C=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10601483
|
Thermo Scientific Maybridge
CC18404CB |
250 mg |
N/A
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10100732
|
Thermo Scientific Maybridge
CC18404DA |
1 g |
N/A
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10747285
|
Thermo Scientific Maybridge
CC18404EB |
25 g |
N/A
|
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
99662-34-7 | |
172.187 | |
PPGRDLZPSDHBIC-UHFFFAOYSA-N | |
2776477 | |
C1=CN(N=C1)C2=CC=C(C=C2)C=O |
C10H8N2O | |
MFCD02681938 | |
4-1h-pyrazol-1-yl benzaldehyde, 4-pyrazol-1-yl-benzaldehyde, 1-4'-formylphenyl pyrazole, 1-4-formylphenyl pyrazole, 4-1h-pyrazol-1-yl benzenecarbaldehyde, 4-pyrazol-1-yl benzaldehyde, 1-4-formylphenyl-1h-pyrazole, benzaldehyde, 4-1h-pyrazol-1-yl, 4-pyrazolylbenzaldehyde, pubchem22713 | |
4-pyrazol-1-ylbenzaldehyde |
Specifications
99662-34-7 | |
100.0 | |
Amber glass bottle | |
MFCD02681938 | |
PPGRDLZPSDHBIC-UHFFFAOYSA-N | |
4-pyrazol-1-ylbenzaldehyde | |
2776477 | |
95% |
94.5 | |
95% | |
C10H8N2O | |
4-1h-pyrazol-1-yl benzaldehyde, 4-pyrazol-1-yl-benzaldehyde, 1-4'-formylphenyl pyrazole, 1-4-formylphenyl pyrazole, 4-1h-pyrazol-1-yl benzenecarbaldehyde, 4-pyrazol-1-yl benzaldehyde, 1-4-formylphenyl-1h-pyrazole, benzaldehyde, 4-1h-pyrazol-1-yl, 4-pyrazolylbenzaldehyde, pubchem22713 | |
C1=CN(N=C1)C2=CC=C(C=C2)C=O | |
172.187 | |
172.19 | |
4-(1H-Pyrazol-1-yl)benzaldehyde |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title