missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Pyrrolidin-1-ylmethyl)aniline, 97%, Thermo Scientific™
£62.10 - £244.00
Chemical Identifiers
CAS | 183365-31-3 |
---|---|
Molecular Formula | C11H16N2 |
Molecular Weight (g/mol) | 176.263 |
InChI Key | RZDBCURLSQNEAD-UHFFFAOYSA-N |
Synonym | 3-pyrrolidin-1-ylmethyl aniline, 3-pyrrolidin-1-ylmethyl-phenylamine, benzenamine, 3-1-pyrrolidinylmethyl, 3-1-pyrrolidinylmethyl aniline, 3-pyrrolidinylmethyl phenylamine, 3-pyrrolidinylmethylaniline, 3-pyrrolidin-1-ylmethyl-aniline, 3-1-pyrrolidinylmethyl benzenamine, 3-pyrrolidin-1-yl-methyl-aniline, 3-pyrrolidin-1-yl methyl aniline |
PubChem CID | 1096407 |
IUPAC Name | 3-(pyrrolidin-1-ylmethyl)aniline |
SMILES | C1CCN(C1)CC2=CC(=CC=C2)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10402464
|
Thermo Scientific Maybridge
CC55714CB |
250 mg |
£62.10
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10419003
|
Thermo Scientific Maybridge
CC55714DA |
1 g |
£82.80
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10235424
|
Thermo Scientific Maybridge
CC55714DE |
5 g |
£244.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
183365-31-3 | |
176.263 | |
3-pyrrolidin-1-ylmethyl aniline, 3-pyrrolidin-1-ylmethyl-phenylamine, benzenamine, 3-1-pyrrolidinylmethyl, 3-1-pyrrolidinylmethyl aniline, 3-pyrrolidinylmethyl phenylamine, 3-pyrrolidinylmethylaniline, 3-pyrrolidin-1-ylmethyl-aniline, 3-1-pyrrolidinylmethyl benzenamine, 3-pyrrolidin-1-yl-methyl-aniline, 3-pyrrolidin-1-yl methyl aniline | |
3-(pyrrolidin-1-ylmethyl)aniline |
C11H16N2 | |
RZDBCURLSQNEAD-UHFFFAOYSA-N | |
1096407 | |
C1CCN(C1)CC2=CC(=CC=C2)N |
Specifications
183365-31-3 | |
C11H16N2 | |
RZDBCURLSQNEAD-UHFFFAOYSA-N | |
3-(pyrrolidin-1-ylmethyl)aniline | |
1096407 | |
97% |
97% | |
3-pyrrolidin-1-ylmethyl aniline, 3-pyrrolidin-1-ylmethyl-phenylamine, benzenamine, 3-1-pyrrolidinylmethyl, 3-1-pyrrolidinylmethyl aniline, 3-pyrrolidinylmethyl phenylamine, 3-pyrrolidinylmethylaniline, 3-pyrrolidin-1-ylmethyl-aniline, 3-1-pyrrolidinylmethyl benzenamine, 3-pyrrolidin-1-yl-methyl-aniline, 3-pyrrolidin-1-yl methyl aniline | |
C1CCN(C1)CC2=CC(=CC=C2)N | |
176.263 | |
176.26 | |
3-(Pyrrolidin-1-ylmethyl)aniline |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title