missing translation for 'onlineSavingsMsg'
Learn More

3-Pyridineacetamide, 99%, Thermo Scientific Chemicals

£38.70 - £156.00

Chemical Identifiers

CAS 3724-16-1
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.154
MDL Number MFCD08236789
InChI Key YDHIMEXEGOCNHU-UHFFFAOYSA-N
Synonym 2-pyridin-3-yl acetamide, 3-pyridineacetamide, pyridine-3-acetamide, 2-3-pyridyl acetamide, 2-pyridine-3-yl-acetamide, 3-pyridine acetamide, 3-pyridylacetamide, acmc-1cs2v, 2-3-pyridinyl acetamide, 2-pyridin-3-yl acetamid
PubChem CID 564767
IUPAC Name 2-pyridin-3-ylacetamide
SMILES C1=CC(=CN=C1)CC(=O)N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11461100
View Documents
Thermo Scientific Alfa Aesar
B25692.06
5 g
£38.70
5g
Please to purchase this item. Need a web account? Register with us today!
11471100
View Documents
Thermo Scientific Alfa Aesar
B25692.14
25 g
£156.00
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3724-16-1
136.154
YDHIMEXEGOCNHU-UHFFFAOYSA-N
564767
C1=CC(=CN=C1)CC(=O)N
C7H8N2O
MFCD08236789
2-pyridin-3-yl acetamide, 3-pyridineacetamide, pyridine-3-acetamide, 2-3-pyridyl acetamide, 2-pyridine-3-yl-acetamide, 3-pyridine acetamide, 3-pyridylacetamide, acmc-1cs2v, 2-3-pyridinyl acetamide, 2-pyridin-3-yl acetamid
2-pyridin-3-ylacetamide

Specifications

3724-16-1
1.17
MFCD08236789
YDHIMEXEGOCNHU-UHFFFAOYSA-N
2-pyridin-3-ylacetamide
564767
99%
122°C to 124°C
C7H8N2O
2-pyridin-3-yl acetamide, 3-pyridineacetamide, pyridine-3-acetamide, 2-3-pyridyl acetamide, 2-pyridine-3-yl-acetamide, 3-pyridine acetamide, 3-pyridylacetamide, acmc-1cs2v, 2-3-pyridinyl acetamide, 2-pyridin-3-yl acetamid
C1=CC(=CN=C1)CC(=O)N
136.154
136.15
3-Pyridineacetamide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only