missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyl-1-propanol, 98%, Thermo Scientific Chemicals

£25.13 - £64.32

Chemical Identifiers

CAS 122-97-4
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.19
MDL Number MFCD00002950
InChI Key VAJVDSVGBWFCLW-UHFFFAOYSA-N
Synonym 3-phenyl-1-propanol, benzenepropanol, hydrocinnamyl alcohol, 3-phenylpropyl alcohol, 3-phenylpropanol, 3-benzenepropanol, phenylpropyl alcohol, hydrocinnamic alcohol, 3-phenyl-n-propanol, 3-hydroxypropyl benzene
PubChem CID 31234
IUPAC Name 3-phenylpropan-1-ol
SMILES C1=CC=C(C=C1)CCCO
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10591411
SDS View Documents
Thermo Scientific Acros
161312500
250 g Glass bottle
£25.13
250g
Please to purchase this item. Need a web account? Register with us today!
10143840
SDS View Documents
Thermo Scientific Acros
161310010
1 kg Glass bottle
£64.32
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

122-97-4
136.19
VAJVDSVGBWFCLW-UHFFFAOYSA-N
31234
C1=CC=C(C=C1)CCCO
C9H12O
MFCD00002950
3-phenyl-1-propanol, benzenepropanol, hydrocinnamyl alcohol, 3-phenylpropyl alcohol, 3-phenylpropanol, 3-benzenepropanol, phenylpropyl alcohol, hydrocinnamic alcohol, 3-phenyl-n-propanol, 3-hydroxypropyl benzene
3-phenylpropan-1-ol

Specifications

122-97-4
100.0
Colorless
235°C
Authentic
C9H12O
1.5250 to 1.527
06, 503
3-phenyl-1-propanol, benzenepropanol, hydrocinnamyl alcohol, 3-phenylpropyl alcohol, 3-phenylpropanol, 3-benzenepropanol, phenylpropyl alcohol, hydrocinnamic alcohol, 3-phenyl-n-propanol, 3-hydroxypropyl benzene
VAJVDSVGBWFCLW-UHFFFAOYSA-N
3-phenylpropan-1-ol
31234
136.19
Liquid
97.5
-18°C
1.0400g/mL
109°C
97.5% min. (GC)
C6H5(CH2)3OH
MFCD00002950
1.04
Solubility in water: 10.3g/L water (20°C). Other solubilities: soluble in methanol,cyclohexane,benzene and,ethylacetate
C1=CC=C(C=C1)CCCO
136.19
20 mPa.s (20°C)
98%
3-Phenyl-1-propanol, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 204-587-6

missing translation for 'rtecsNumber' : UB8970000

missing translation for 'tsca' : TSCA

Product Identifier
  • 3-Phenyl-1-propanol

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.