missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Morpholin-4-ylmethyl)benzoic acid hydrochloride hydrate, 97%, Thermo Scientific™
£29.90 - £74.80
Chemical Identifiers
CAS | 137605-80-2 |
---|---|
Molecular Formula | C12H16ClNO3 |
Molecular Weight (g/mol) | 257.71 |
MDL Number | MFCD09064949 |
InChI Key | UOMMCPQFAXOMMO-UHFFFAOYSA-N |
Synonym | 3-morpholin-4-ylmethyl benzoic acid hydrochloride, 3-morpholinomethyl benzoic acid hydrochloride, 3-morpholin-4-ylmethyl benzoic acid, chloride, 3-morpholin-4-ylmethyl benzoic acid,hydrochloride, 3-morpholin-4-yl methyl benzoic acid hydrochloride, 3-morpholin-4-yl methyl benzoic acid-hydrogen chloride 1/1 |
PubChem CID | 24229486 |
IUPAC Name | 3-(morpholin-4-ylmethyl)benzoic acid;hydrochloride |
SMILES | Cl.OC(=O)C1=CC(CN2CCOCC2)=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10527663
|
Thermo Scientific Maybridge
CC40151CB |
250 mg |
£29.90
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10729983
|
Thermo Scientific Maybridge
CC40151DA |
1 g |
£74.80
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
137605-80-2 | |
257.71 | |
UOMMCPQFAXOMMO-UHFFFAOYSA-N | |
24229486 | |
Cl.OC(=O)C1=CC(CN2CCOCC2)=CC=C1 |
C12H16ClNO3 | |
MFCD09064949 | |
3-morpholin-4-ylmethyl benzoic acid hydrochloride, 3-morpholinomethyl benzoic acid hydrochloride, 3-morpholin-4-ylmethyl benzoic acid, chloride, 3-morpholin-4-ylmethyl benzoic acid,hydrochloride, 3-morpholin-4-yl methyl benzoic acid hydrochloride, 3-morpholin-4-yl methyl benzoic acid-hydrogen chloride 1/1 | |
3-(morpholin-4-ylmethyl)benzoic acid;hydrochloride |
Specifications
137605-80-2 | |
100.0 | |
C12H16ClNO3 | |
3-morpholin-4-ylmethyl benzoic acid hydrochloride, 3-morpholinomethyl benzoic acid hydrochloride, 3-morpholin-4-ylmethyl benzoic acid, chloride, 3-morpholin-4-ylmethyl benzoic acid,hydrochloride, 3-morpholin-4-yl methyl benzoic acid hydrochloride, 3-morpholin-4-yl methyl benzoic acid-hydrogen chloride 1/1 | |
Cl.OC(=O)C1=CC(CN2CCOCC2)=CC=C1 | |
257.71 | |
275.74 | |
3-(Morpholin-4-ylmethyl)benzoic acid hydrochloride hydrate |
96.5 | |
97% | |
MFCD09064949 | |
UOMMCPQFAXOMMO-UHFFFAOYSA-N | |
3-(morpholin-4-ylmethyl)benzoic acid;hydrochloride | |
24229486 | |
97% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing
Warning
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title