Learn More
3-Hydroxypiperidine hydrochloride, 98+%, Thermo Scientific™
£21.20 - £98.70
Chemical Identifiers
CAS | 64051-79-2 |
---|---|
Molecular Formula | C5H12ClNO |
Molecular Weight (g/mol) | 137.607 |
MDL Number | MFCD00012772 |
InChI Key | VLECDMDGMKPUSK-UHFFFAOYSA-N |
Synonym | 3-hydroxypiperidine hydrochloride, piperidin-3-ol hydrochloride, 3-hydroxypiperidine hcl, 3-piperidinol hydrochloride, piperidine-3-ol hydrochloride, 3-hydroxy-piperidine hydrochloride, 3-piperidinol, hydrochloride, piperidine-3-ol hcl, 3-hydroxypiperidine.hcl, 3-hydroxypiperidinehydrochloride |
PubChem CID | 2723962 |
IUPAC Name | piperidin-3-ol;hydrochloride |
SMILES | C1CC(CNC1)O.Cl |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications3-Hydroxypiperidine hydrochloride is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in methanol.
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Chemical Identifiers
64051-79-2 | |
137.607 | |
VLECDMDGMKPUSK-UHFFFAOYSA-N | |
2723962 | |
C1CC(CNC1)O.Cl |
C5H12ClNO | |
MFCD00012772 | |
3-hydroxypiperidine hydrochloride, piperidin-3-ol hydrochloride, 3-hydroxypiperidine hcl, 3-piperidinol hydrochloride, piperidine-3-ol hydrochloride, 3-hydroxy-piperidine hydrochloride, 3-piperidinol, hydrochloride, piperidine-3-ol hcl, 3-hydroxypiperidine.hcl, 3-hydroxypiperidinehydrochloride | |
piperidin-3-ol;hydrochloride |
Specifications
64051-79-2 | |
MFCD00012772 | |
Soluble in methanol. | |
C1CC(CNC1)O.Cl | |
137.607 | |
137.61 | |
3-Hydroxypiperidine hydrochloride |
C5H12ClNO | |
3-hydroxypiperidine hydrochloride, piperidin-3-ol hydrochloride, 3-hydroxypiperidine hcl, 3-piperidinol hydrochloride, piperidine-3-ol hydrochloride, 3-hydroxy-piperidine hydrochloride, 3-piperidinol, hydrochloride, piperidine-3-ol hcl, 3-hydroxypiperidine.hcl, 3-hydroxypiperidinehydrochloride | |
VLECDMDGMKPUSK-UHFFFAOYSA-N | |
piperidin-3-ol;hydrochloride | |
2723962 | |
≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 264-644-6
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures