missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Dimethylamino-1-propanol, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Dimethylamino-1-propanol, is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
Solubility
It is miscible with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
3-Dimethylamino-1-propanol, is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
Solubility
It is miscible with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
Melting Point | -35°C |
Density | 0.88 |
Boiling Point | 162°C to 164°C |
Flash Point | 54°C (129°F) |
Linear Formula | (CH3)2N(CH2)3OH |
Refractive Index | 1.436 |
Quantity | 100 mL |
UN Number | UN2734 |
Beilstein | 1732032 |
Sensitivity | Air Sensitive |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'