missing translation for 'onlineSavingsMsg'
Learn More

3-Di-n-propylamino-1-propyne, 98%, Thermo Scientific™

£21.20 - £51.50

Chemical Identifiers

CAS 6323-79-1
Molecular Formula C9H17N
Molecular Weight (g/mol) 139.242
MDL Number MFCD00041626
InChI Key ATTMKPWGJILFFV-UHFFFAOYSA-N
Synonym 3-di-n-propylamino-1-propyne, n,n-dipropyl-2-propyn-1-amine, propargyldipropylamine, n,n-dipropylprop-2-yn-1-amine, 3-dipropylamino-1-propyne, 2-propyn-1-amine, n,n-dipropyl, prop-2-ynyldipropylamine, dipropylpropargylamine, propargyl di-n-propyl amine, n,n-dipropyl-2-propynylamine
PubChem CID 138724
IUPAC Name N-propyl-N-prop-2-ynylpropan-1-amine
SMILES CCCN(CCC)CC#C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11352537
View Documents
Thermo Scientific Alfa Aesar
L11633.03
1 g
£21.20
1g
Please to purchase this item. Need a web account? Register with us today!
11362537
View Documents
Thermo Scientific Alfa Aesar
L11633.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

6323-79-1
139.242
ATTMKPWGJILFFV-UHFFFAOYSA-N
138724
CCCN(CCC)CC#C
C9H17N
MFCD00041626
3-di-n-propylamino-1-propyne, n,n-dipropyl-2-propyn-1-amine, propargyldipropylamine, n,n-dipropylprop-2-yn-1-amine, 3-dipropylamino-1-propyne, 2-propyn-1-amine, n,n-dipropyl, prop-2-ynyldipropylamine, dipropylpropargylamine, propargyl di-n-propyl amine, n,n-dipropyl-2-propynylamine
N-propyl-N-prop-2-ynylpropan-1-amine

Specifications

6323-79-1
157°C to 159°C
Amine-like
1.435
UN2733
3-di-n-propylamino-1-propyne, n,n-dipropyl-2-propyn-1-amine, propargyldipropylamine, n,n-dipropylprop-2-yn-1-amine, 3-dipropylamino-1-propyne, 2-propyn-1-amine, n,n-dipropyl, prop-2-ynyldipropylamine, dipropylpropargylamine, propargyl di-n-propyl amine, n,n-dipropyl-2-propynylamine
CCCN(CCC)CC#C
139.242
139.24
3-Di-n-propylamino-1-propyne
0.828
55°C (131°F)
C9H17N
MFCD00041626
1746300
ATTMKPWGJILFFV-UHFFFAOYSA-N
N-propyl-N-prop-2-ynylpropan-1-amine
138724
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H226-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications