missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclopropyl-1-methyl-1H-pyrazol-5-amine, 97%, Thermo Scientific™

Product Code. 10553022
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 118430-74-3
Molecular Formula C7H11N3
Molecular Weight (g/mol) 137.19
MDL Number MFCD00067984
InChI Key YWHWPIRLFHZSFS-UHFFFAOYSA-N
PubChem CID 2735311
SMILES CN1N=C(C=C1N)C1CC1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Synonym 3-cyclopropyl-1-methyl-1h-pyrazol-5-amine, 1h-pyrazol-5-amine, 3-cyclopropyl-1-methyl, 5-amino-3-cyclopropyl-1-methylpyrazole, 5-amino-3-cyclopropyl-1-methyl-1h-pyrazole, 5-cyclopropyl-2-methyl-2h-pyrazol-3-ylamine, 1h-pyrazol-5-amine,3-cyclopropyl-1-methyl, 3-cyclopropyl-1-methylpyrazole-5-ylamine, acmc-1cbj7, 5-amino-3-cyclopropyl-1-methyl pyrazole, 5-cyclopropyl-2-methyl-pyrazole-3-amine
IUPAC Name 3-cyclopropyl-1-methyl-1H-pyrazol-5-amine
Formula Weight 137.18
Percent Purity 97%
Chemical Name or Material 3-Cyclopropyl-1-methyl-1H-pyrazol-5-amine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.