missing translation for 'onlineSavingsMsg'
Learn More

3-(Cyclopentylcarbamoyl)benzeneboronic acid, 98%, Thermo Scientific™

Product Code. 15440406
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850567-24-7
Molecular Formula C12H16BNO3
Molecular Weight (g/mol) 233.074
MDL Number MFCD04115695
InChI Key KIOBFDGZJVJNBP-UHFFFAOYSA-N
Synonym 3-cyclopentylaminocarbonyl phenylboronic acid, 3-cyclopentylcarbamoyl phenyl boronic acid, 3-cyclopentylcarbamoyl phenylboronic acid, 3-cyclopentylaminocarbonyl benzeneboronic acid, pubchem16391, acmc-209q0d, 3-cyclopentylcarbamoyl benzeneboronic acid, n-cyclopentyl-3-boronobenzamide
PubChem CID 3775808
IUPAC Name [3-(cyclopentylcarbamoyl)phenyl]boronic acid
SMILES B(C1=CC(=CC=C1)C(=O)NC2CCCC2)(O)O

Specifications

Quantity 250 mg
Formula Weight 233.07
Percent Purity 98%
Chemical Name or Material 3-(Cyclopentylcarbamoyl)benzeneboronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.