Learn More
3-Chloropyridine, 99%, Thermo Scientific Chemicals
£36.68 - £102.51
Chemical Identifiers
CAS | 626-60-8 |
---|---|
Molecular Formula | C5H4ClN |
Molecular Weight (g/mol) | 113.54 |
MDL Number | MFCD00006375 |
InChI Key | PWRBCZZQRRPXAB-UHFFFAOYSA-N |
Synonym | pyridine, 3-chloro, m-chloropyridine, unii-1m13huc1p4, ccris 1715, chloropyridine, b-chloropyridine, 3-chloro pyridine, 3-chloro-pyridine, 3-chloropyridine, 3-chloranylpyridine |
PubChem CID | 12287 |
IUPAC Name | 3-chloropyridine |
SMILES | ClC1=CC=CN=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10162230
|
Thermo Scientific Acros
110020250 |
25 mL | Glass bottle |
£36.68
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10494935
|
Thermo Scientific Acros
110021000 |
100 mL | Glass bottle |
£102.51
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
626-60-8 | |
113.54 | |
PWRBCZZQRRPXAB-UHFFFAOYSA-N | |
12287 | |
ClC1=CC=CN=C1 |
C5H4ClN | |
MFCD00006375 | |
pyridine, 3-chloro, m-chloropyridine, unii-1m13huc1p4, ccris 1715, chloropyridine, b-chloropyridine, 3-chloro pyridine, 3-chloro-pyridine, 3-chloropyridine, 3-chloranylpyridine | |
3-chloropyridine |
Specifications
626-60-8 | |
100.0 | |
1.1900g/mL | |
66°C | |
98.5% min. (GC) | |
(C5H4N)Cl | |
MFCD00006375 | |
1.19 | |
Solubility in water: 10g/L (20°C) | |
ClC1=CC=CN=C1 | |
113.54 | |
113.55 | |
Liquid |
98.5 | |
Colorless to Yellow | |
147.0°C to 150.0°C | |
Authentic | |
C5H4ClN | |
1.5310 to 1.5340 | |
20, 230 | |
pyridine, 3-chloro, m-chloropyridine, unii-1m13huc1p4, ccris 1715, chloropyridine, b-chloropyridine, 3-chloro pyridine, 3-chloro-pyridine, 3-chloropyridine, 3-chloranylpyridine | |
PWRBCZZQRRPXAB-UHFFFAOYSA-N | |
3-chloropyridine | |
12287 | |
99% | |
3-Chloropyridine |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physicia
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 210-955-7
- 3-Chloropyridine
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only