Learn More
3-Chloro-1-propanol, 98%, stab., Thermo Scientific Chemicals
£24.72 - £213.00
Chemical Identifiers
CAS | 627-30-5 |
---|---|
Molecular Formula | C3H7ClO |
Molecular Weight (g/mol) | 94.538 |
MDL Number | MFCD00002943 |
InChI Key | LAMUXTNQCICZQX-UHFFFAOYSA-N |
Synonym | 3-chloro-1-propanol, 3-chloropropanol, 1-propanol, 3-chloro, 1-chloro-3-hydroxypropane, trimethylene chlorohydrin, 3-chloro-l-propanol, 3-chlorpropan-1-ol, 3-choro-1-propanol, 1-chloro-3-propanol, 3-chloropropanol-1 |
PubChem CID | 12313 |
IUPAC Name | 3-chloropropan-1-ol |
SMILES | C(CO)CCl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11399499
|
Thermo Scientific Alfa Aesar
A16871.14 |
25 g |
£24.72
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11492898
|
Thermo Scientific Alfa Aesar
A16871.22 |
100 g |
£52.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11402908
|
Thermo Scientific Alfa Aesar
A16871.36 |
500 g |
£213.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a pharmaceutical intermediate.
Solubility
Soluble in water at 20°C 300g/L.
Notes
Store away from oxidizing agents and bases. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
Chemical Identifiers
627-30-5 | |
94.538 | |
LAMUXTNQCICZQX-UHFFFAOYSA-N | |
12313 | |
C(CO)CCl |
C3H7ClO | |
MFCD00002943 | |
3-chloro-1-propanol, 3-chloropropanol, 1-propanol, 3-chloro, 1-chloro-3-hydroxypropane, trimethylene chlorohydrin, 3-chloro-l-propanol, 3-chlorpropan-1-ol, 3-choro-1-propanol, 1-chloro-3-propanol, 3-chloropropanol-1 | |
3-chloropropan-1-ol |
Specifications
627-30-5 | |
1.131 | |
160°C to 162°C | |
Pleasant | |
1.445 | |
UN2849 | |
3-chloro-1-propanol, 3-chloropropanol, 1-propanol, 3-chloro, 1-chloro-3-hydroxypropane, trimethylene chlorohydrin, 3-chloro-l-propanol, 3-chlorpropan-1-ol, 3-choro-1-propanol, 1-chloro-3-propanol, 3-chloropropanol-1 | |
LAMUXTNQCICZQX-UHFFFAOYSA-N | |
3-chloropropan-1-ol | |
12313 | |
98% |
-20°C | |
3 to 4 | |
73°C (163°F) | |
C3H7ClO | |
MFCD00002943 | |
773655 | |
Soluble in water at 20°C 300g/L. | |
C(CO)CCl | |
94.538 | |
94.54 | |
3-Chloro-1-propanol |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: 3-CHLOROPROPANOL-1
missing translation for 'einecsNumber' : 210-992-9
missing translation for 'rtecsNumber' : UA8930000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only