Learn More
3-Bromo-N-methylaniline, 97%, Thermo Scientific Chemicals
£27.10 - £443.00
Chemical Identifiers
CAS | 66584-32-5 |
---|---|
Molecular Formula | C7H8BrN |
Molecular Weight (g/mol) | 186.052 |
MDL Number | MFCD05664376 |
InChI Key | HKOSFZXROYRVJT-UHFFFAOYSA-N |
Synonym | n-methyl-3-bromoaniline, 3-bromo-n-methyl aniline, benzenamine, 3-bromo-n-methyl, acmc-20an89, 3-bromophenyl-methylamine, 3-bromo-phenyl-methyl-amine, 3-bromo-n-methylaniline |
PubChem CID | 7018299 |
IUPAC Name | 3-bromo-N-methylaniline |
SMILES | CNC1=CC(=CC=C1)Br |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
66584-32-5 | |
186.052 | |
HKOSFZXROYRVJT-UHFFFAOYSA-N | |
7018299 | |
CNC1=CC(=CC=C1)Br |
C7H8BrN | |
MFCD05664376 | |
n-methyl-3-bromoaniline, 3-bromo-n-methyl aniline, benzenamine, 3-bromo-n-methyl, acmc-20an89, 3-bromophenyl-methylamine, 3-bromo-phenyl-methyl-amine, 3-bromo-n-methylaniline | |
3-bromo-N-methylaniline |
Specifications
66584-32-5 | |
1.461 g/mL | |
>110°C (230°F) | |
C7H8BrN | |
MFCD05664376 | |
HKOSFZXROYRVJT-UHFFFAOYSA-N | |
3-bromo-N-methylaniline | |
7018299 | |
97% | |
3-Bromo-N-methylaniline |
Colorless to Yellow | |
253°C | |
Unpleasant | |
1.621 | |
n-methyl-3-bromoaniline, 3-bromo-n-methyl aniline, benzenamine, 3-bromo-n-methyl, acmc-20an89, 3-bromophenyl-methylamine, 3-bromo-phenyl-methyl-amine, 3-bromo-n-methylaniline | |
CNC1=CC(=CC=C1)Br | |
186.052 | |
186.05 | |
Liquid |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.