missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopropyltrimethoxysilane, 95%, Thermo Scientific Chemicals

Product Code. 10589453
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13822-56-5
Molecular Formula C6H17NO3Si
Molecular Weight (g/mol) 179.29
MDL Number MFCD00008206
InChI Key SJECZPVISLOESU-UHFFFAOYSA-N
Synonym 3-aminopropyltrimethoxysilane, 3-aminopropyl trimethoxysilane, 3-trimethoxysilyl-1-propanamine, 3-trimethoxysilyl propan-1-amine, 3-trimethoxysilyl propylamine, 1-propanamine, 3-trimethoxysilyl, silane sc 3900, n-trimethoxysilylpropyl amine, propylamine, 3-trimethoxysilyl, unii-7mry0c9sb6
PubChem CID 83756
IUPAC Name 3-trimethoxysilylpropan-1-amine
SMILES CO[Si](CCCN)(OC)OC

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless
Density 1.0100g/mL
Boiling Point 194.0°C
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula NH2(CH2)3Si(OCH3)3
Refractive Index 1.4230 to 1.4250
Quantity 500 mL
Specific Gravity 1.01
Solubility Information Solubility in water: reacts with water.
Formula Weight 179.29
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3-Aminopropyltrimethoxysilane
Show More Show Less
compliance-icons
Product Identifier
  • 3-Aminopropyltrimethoxysilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 3-Aminopropyltrimethoxysilane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'