Learn More
3-Amino-1-propanol, 99%, Thermo Scientific Chemicals
£23.82 - £55.28
Chemical Identifiers
CAS | 156-87-6 |
---|---|
Molecular Formula | C3H9NO |
Molecular Weight (g/mol) | 75.111 |
MDL Number | MFCD00008223 |
InChI Key | WUGQZFFCHPXWKQ-UHFFFAOYSA-N |
Synonym | 3-aminopropanol, 3-amino-1-propanol, propanolamine, 1-propanol, 3-amino, 3-propanolamine, 1-amino-3-propanol, 3-hydroxypropylamine, 3-aminopropyl alcohol, 1,3-propanolamine, beta-alaninol |
PubChem CID | 9086 |
IUPAC Name | 3-aminopropan-1-ol |
SMILES | C(CN)CO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11496226
|
Thermo Scientific Alfa Aesar
B23041.22 |
100 g |
£23.82
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11406236
|
Thermo Scientific Alfa Aesar
B23041.36 |
500 g |
£55.28
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-Amino-1-propanol is used in the production of anionic emulsifiers, nonionic polyethylene emulsions. Its salts such as hydrochloride, nitrate, sulfate, phosphate are used for the balanced curing action with selected textile resins. It is used as a starting material in the preparation of beta-lactam antibiotics, humectants for foods and cosmetics. It acts as a corrosion inhibitor and finds application in water treatment, metal treatment and absorption of carbon dioxide gas.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Amino-1-propanol is used in the production of anionic emulsifiers, nonionic polyethylene emulsions. Its salts such as hydrochloride, nitrate, sulfate, phosphate are used for the balanced curing action with selected textile resins. It is used as a starting material in the preparation of beta-lactam antibiotics, humectants for foods and cosmetics. It acts as a corrosion inhibitor and finds application in water treatment, metal treatment and absorption of carbon dioxide gas.
Solubility
Miscible with water, alcohol, ether, acetone and chloroform.
Notes
Hygroscopic. Incompatible withstrong oxidizing agents.
Chemical Identifiers
156-87-6 | |
75.111 | |
WUGQZFFCHPXWKQ-UHFFFAOYSA-N | |
9086 | |
C(CN)CO |
C3H9NO | |
MFCD00008223 | |
3-aminopropanol, 3-amino-1-propanol, propanolamine, 1-propanol, 3-amino, 3-propanolamine, 1-amino-3-propanol, 3-hydroxypropylamine, 3-aminopropyl alcohol, 1,3-propanolamine, beta-alaninol | |
3-aminopropan-1-ol |
Specifications
156-87-6 | |
0.983 | |
101°C (214°F) | |
C3H9NO | |
MFCD00008223 | |
741855 | |
3-aminopropanol, 3-amino-1-propanol, propanolamine, 1-propanol, 3-amino, 3-propanolamine, 1-amino-3-propanol, 3-hydroxypropylamine, 3-aminopropyl alcohol, 1,3-propanolamine, beta-alaninol | |
WUGQZFFCHPXWKQ-UHFFFAOYSA-N | |
3-aminopropan-1-ol | |
9086 | |
99% |
11°C | |
184°C to 187°C | |
Amine-like | |
1.461 | |
UN2735 | |
Hygroscopic | |
Miscible with water,alcohol,ether,acetone and chloroform. | |
C(CN)CO | |
75.111 | |
75.11 | |
3-Amino-1-propanol |
Safety and Handling
GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H332-H311-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 205-864-4
missing translation for 'rtecsNumber' : UA5600000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only