Learn More
3-Acetylthiophene, 98%, Thermo Scientific Chemicals
£70.35 - £1141.68
Chemical Identifiers
CAS | 1468-83-3 |
---|---|
Molecular Formula | C6H6OS |
Molecular Weight (g/mol) | 126.17 |
MDL Number | MFCD00005468 |
InChI Key | RNIDWJDZNNVFDY-UHFFFAOYSA-N |
Synonym | 3-acetylthiophene, methyl 3-thienyl ketone, 1-thiophen-3-yl ethan-1-one, 1-thiophen-3-yl ethanone, 1-3-thienyl ethanone, ethanone, 1-3-thienyl, ketone, methyl 3-thienyl, 3-acetyl thiophene, 1-thien-3-ylethanone, methyl-3-thienyl ketone |
PubChem CID | 15116 |
IUPAC Name | 1-thiophen-3-ylethanone |
SMILES | CC(=O)C1=CSC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11408943
|
Thermo Scientific Alfa Aesar
A11078.09 |
10 g |
£70.35
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11418943
|
Thermo Scientific Alfa Aesar
A11078.18 |
50 g |
£229.14
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11428943
|
Thermo Scientific Alfa Aesar
A11078.30 |
250 g |
£1141.68
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1468-83-3 | |
126.17 | |
RNIDWJDZNNVFDY-UHFFFAOYSA-N | |
15116 | |
CC(=O)C1=CSC=C1 |
C6H6OS | |
MFCD00005468 | |
3-acetylthiophene, methyl 3-thienyl ketone, 1-thiophen-3-yl ethan-1-one, 1-thiophen-3-yl ethanone, 1-3-thienyl ethanone, ethanone, 1-3-thienyl, ketone, methyl 3-thienyl, 3-acetyl thiophene, 1-thien-3-ylethanone, methyl-3-thienyl ketone | |
1-thiophen-3-ylethanone |
Specifications
1468-83-3 | |
208°C to 210°C | |
MFCD00005468 | |
107241 | |
3-acetylthiophene, methyl 3-thienyl ketone, 1-thiophen-3-yl ethan-1-one, 1-thiophen-3-yl ethanone, 1-3-thienyl ethanone, ethanone, 1-3-thienyl, ketone, methyl 3-thienyl, 3-acetyl thiophene, 1-thien-3-ylethanone, methyl-3-thienyl ketone | |
CC(=O)C1=CSC=C1 | |
126.17 | |
126.18 | |
3-Acetylthiophene |
58°C to 61°C | |
C6H6OS | |
UN2811 | |
Light sensitive | |
RNIDWJDZNNVFDY-UHFFFAOYSA-N | |
1-thiophen-3-ylethanone | |
15116 | |
98% |
Safety and Handling
GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 215-996-4
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.