Learn More
3',5'-Bis(trifluoromethyl)acetophenone, 98%, Thermo Scientific Chemicals
£19.40 - £197.00
Chemical Identifiers
CAS | 30071-93-3 |
---|---|
Molecular Formula | C10H6F6O |
Molecular Weight (g/mol) | 256.15 |
MDL Number | MFCD00009910 |
InChI Key | MCYCSIKSZLARBD-UHFFFAOYSA-N |
Synonym | 3',5'-bis trifluoromethyl acetophenone, 3,5-bis trifluoromethyl acetophenone, 1-3,5-bis trifluoromethyl phenyl ethanone, 1-3,5-bis trifluoromethyl phenyl ethan-1-one, mbt-ac, 1-3,5-di trifluoromethyl phenyl ethan-1-one, 3,5-ditrifluoromethylacetophenone, 3',5'-bis-trifluoromethylacetophenone |
PubChem CID | 121616 |
SMILES | CC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11484123
|
Thermo Scientific Alfa Aesar
A10341.03 |
1 g |
£19.40
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11494123
|
Thermo Scientific Alfa Aesar
A10341.06 |
5 g |
£51.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11404133
|
Thermo Scientific Alfa Aesar
A10341.14 |
25 g |
£197.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
30071-93-3 | |
256.15 | |
MCYCSIKSZLARBD-UHFFFAOYSA-N | |
121616 |
C10H6F6O | |
MFCD00009910 | |
3',5'-bis trifluoromethyl acetophenone, 3,5-bis trifluoromethyl acetophenone, 1-3,5-bis trifluoromethyl phenyl ethanone, 1-3,5-bis trifluoromethyl phenyl ethan-1-one, mbt-ac, 1-3,5-di trifluoromethyl phenyl ethan-1-one, 3,5-ditrifluoromethylacetophenone, 3',5'-bis-trifluoromethylacetophenone | |
CC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F |
Specifications
30071-93-3 | |
85°C (8 mmHg) | |
C10H6F6O | |
MFCD00009910 | |
3',5'-bis trifluoromethyl acetophenone, 3,5-bis trifluoromethyl acetophenone, 1-3,5-bis trifluoromethyl phenyl ethanone, 1-3,5-bis trifluoromethyl phenyl ethan-1-one, mbt-ac, 1-3,5-di trifluoromethyl phenyl ethan-1-one, 3,5-ditrifluoromethylacetophenone, 3',5'-bis-trifluoromethylacetophenone | |
CC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F | |
256.15 | |
256.15 | |
3',5'-Bis (trifluoromethyl)acetophenone |
1.422 | |
82°C (179°F) | |
1.423 | |
2384789 | |
MCYCSIKSZLARBD-UHFFFAOYSA-N | |
1-[3,5-bis(trifluoromethyl)phenyl]ethan-1-one | |
121616 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302+H312+H332-H315-H319-H335
missing translation for 'einecsNumber' : 250-023-7
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only