missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-diethoxycyclobut-3-ene-1,2-dione, Thermo Scientific™
£90.90 - £294.00
Chemical Identifiers
CAS | 5231-87-8 |
---|---|
Molecular Formula | C8H10O4 |
Molecular Weight (g/mol) | 170.164 |
InChI Key | DFSFLZCLKYZYRD-UHFFFAOYSA-N |
Synonym | 3,4-diethoxy-3-cyclobutene-1,2-dione, diethyl squarate, squaric acid diethyl ester, 3-cyclobutene-1,2-dione, 3,4-diethoxy, diethoxycyclobutenedione, diethoxycyclobut-3-ene-1,2-dione, 1,2-diethoxycyclobutenedione, 3,4-diethoxy-cyclobut-3-ene-1,2-dione, pubchem9741, acmc-209kyn |
PubChem CID | 123228 |
IUPAC Name | 3,4-diethoxycyclobut-3-ene-1,2-dione |
SMILES | CCOC1=C(C(=O)C1=O)OCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10768243
|
Thermo Scientific Maybridge
RDP00117EA |
10 g | Amber Glass Bottle |
£90.90
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10372015
|
Thermo Scientific Maybridge
RDP00117EB |
25 g | Amber Glass Bottle |
£176.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10497554
|
Thermo Scientific Maybridge
RDP00117FA |
100 g | Amber Glass Bottle |
£294.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
5231-87-8 | |
170.164 | |
3,4-diethoxy-3-cyclobutene-1,2-dione, diethyl squarate, squaric acid diethyl ester, 3-cyclobutene-1,2-dione, 3,4-diethoxy, diethoxycyclobutenedione, diethoxycyclobut-3-ene-1,2-dione, 1,2-diethoxycyclobutenedione, 3,4-diethoxy-cyclobut-3-ene-1,2-dione, pubchem9741, acmc-209kyn | |
3,4-diethoxycyclobut-3-ene-1,2-dione |
C8H10O4 | |
DFSFLZCLKYZYRD-UHFFFAOYSA-N | |
123228 | |
CCOC1=C(C(=O)C1=O)OCC |
Specifications
5231-87-8 | |
C8H10O4 | |
DFSFLZCLKYZYRD-UHFFFAOYSA-N | |
3,4-diethoxycyclobut-3-ene-1,2-dione | |
123228 | |
97% |
97% | |
3,4-diethoxy-3-cyclobutene-1,2-dione, diethyl squarate, squaric acid diethyl ester, 3-cyclobutene-1,2-dione, 3,4-diethoxy, diethoxycyclobutenedione, diethoxycyclobut-3-ene-1,2-dione, 1,2-diethoxycyclobutenedione, 3,4-diethoxy-cyclobut-3-ene-1,2-dione, pubchem9741, acmc-209kyn | |
CCOC1=C(C(=O)C1=O)OCC | |
170.164 | |
170.16 | |
3, 4-Diethoxycyclobut-3-ene-1, 2-dione |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title