missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(4,5-Dimethyl-2-thazolyl)-2,5-diphenyl-2H-tetrazolium bromide, ∽98%, MP Biomedicals™
£130.00 - £865.00
Chemical Identifiers
CAS | 298-93-1 |
---|---|
Molecular Formula | C18H16BrN5S |
Molecular Weight (g/mol) | 414.33 |
MDL Number | MFCD00011964,MFCD00066662 |
InChI Key | AZKSAVLVSZKNRD-UHFFFAOYSA-M |
Synonym | thiazolyl blue, thiazolyl blue tetrazolium bromide, mmt tetrazolium, methylthiazoletetrazolium, mtt, thiazolyl blue monotetrazolium, thiazole blue, 3-4,5-dimethylthiazol-2-yl-2,5-diphenyltetrazolium bromide, mtt van, unii-euy85h477i |
PubChem CID | 64965 |
ChEBI | CHEBI:53233 |
SMILES | [Br-].CC1=C(C)N=C(S1)[N+]1=NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11399079
|
MP Biomedicals
0210222701 |
1 g |
£130.00
21g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11319089
|
MP Biomedicals
0210222710 |
10 g |
£865.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
298-93-1 | |
414.33 | |
AZKSAVLVSZKNRD-UHFFFAOYSA-M | |
64965 | |
[Br-].CC1=C(C)N=C(S1)[N+]1=NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1 |
C18H16BrN5S | |
MFCD00011964,MFCD00066662 | |
thiazolyl blue, thiazolyl blue tetrazolium bromide, mmt tetrazolium, methylthiazoletetrazolium, mtt, thiazolyl blue monotetrazolium, thiazole blue, 3-4,5-dimethylthiazol-2-yl-2,5-diphenyltetrazolium bromide, mtt van, unii-euy85h477i | |
CHEBI:53233 |
Specifications
298-93-1 | |
C18H16BrN5S | |
thiazolyl blue, thiazolyl blue tetrazolium bromide, mmt tetrazolium, methylthiazoletetrazolium, mtt, thiazolyl blue monotetrazolium, thiazole blue, 3-4,5-dimethylthiazol-2-yl-2,5-diphenyltetrazolium bromide, mtt van, unii-euy85h477i | |
[Br-].CC1=C(C)N=C(S1)[N+]1=NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1 | |
414.33 | |
CHEBI:53233 | |
∼98% | |
3-(4, 5-Dimethyl-2-thazolyl)-2, 5-diphenyl-2H-tetrazolium bromide |
Orange-Yellow | |
MFCD00011964,MFCD00066662 | |
AZKSAVLVSZKNRD-UHFFFAOYSA-M | |
2-(3,5-diphenyltetrazol-2-ium-2-yl)-4,5-dimethyl-1,3-thiazole;bromide | |
64965 | |
414.32 | |
Crystalline Powder |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 3-(4,5-Dimethylthiazolyl-2)-2,5-diphenyl Tetrazolium Bromide
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H341-Suspected of causing genetic defects.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 3-(4,5-Dimethylthiazolyl-2)-2,5-diphenylTetrazolium Bromide
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3-(4,5-Dimethyl-2-thazolyl)-2,5-diphenyl-2H-tetrazolium bromide, ∽98%, MP Biomedicals™