missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethylpiperidine, 95%, Thermo Scientific Chemicals

Product Code. 11349969
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1193-12-0
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.20
MDL Number MFCD00005995
InChI Key CDODDZJCEADUQQ-UHFFFAOYSA-N
PubChem CID 70942
IUPAC Name 3,3-dimethylpiperidine
SMILES CC1(C)CCCNC1

Specifications

Density 0.81
Boiling Point 134°C to 136°C
Flash Point 26°C (79°F)
Odor Amine-like
Refractive Index 1.4475
Quantity 1 g
UN Number UN2733
Sensitivity Air Sensitive
Synonym piperidine, 3,3-dimethyl, 3,3-dimethyl-piperidine, 3,3-dimethyl piperidine, pubchem10316, acmc-1bo7w, 3,3-dimethylpiperdine
Formula Weight 113.2
Percent Purity 95%
Chemical Name or Material 3,3-Dimethylpiperidine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.