missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethylcyclohexanone, 97%, Thermo Scientific Chemicals

Product Code. 11347559
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

An interest in the boll weevil sex attractants prompted an investigation into alternative syntheses of the starting material, 3, 3-dimethylcyclohexanone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An interest in the boll weevil sex attractants prompted an investigation into alternative syntheses of the starting material, 3, 3-dimethylcyclohexanone.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2979-19-3
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.20
MDL Number MFCD00040178
InChI Key ZVJQBBYAVPAFLX-UHFFFAOYSA-N
Synonym 3,3-dimethylcyclohexanone, cyclohexanone, 3,3-dimethyl, 3,3-dimethylcyclohexane-1-one, 3,3-dimethyl-cyclohexanone, 3,3-dimethylcyclohexanon, acmc-209hb2, 3,3-dimethyl cyclohexanone, ksc497k3f, 3,3-dimethyl-cyclohexan-1-one, zvjqbbyavpaflx-uhfffaoysa
PubChem CID 76322
IUPAC Name 3,3-dimethylcyclohexan-1-one
SMILES CC1(C)CCCC(=O)C1

Specifications

Density 0.91
Boiling Point 175°C
Flash Point 59°C (138°F)
Refractive Index 1.447
Quantity 1 g
UN Number UN1224
Solubility Information Insoluble in water.
Formula Weight 126.2
Percent Purity 97%
Chemical Name or Material 3,3-Dimethylcyclohexanone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'