missing translation for 'onlineSavingsMsg'
Learn More

[3-(2-Furyl)phenyl]methanol, ≥97%, Thermo Scientific™

£95.50 - £289.00

Chemical Identifiers

CAS 89929-93-1
Molecular Formula C11H10O2
Molecular Weight (g/mol) 174.199
MDL Number MFCD06203100
InChI Key SBTRFADZILHXNG-UHFFFAOYSA-N
Synonym 3-2-furyl phenyl methanol, 3-furan-2-yl phenyl methanol, 3-2-furyl benzyl alcohol, 3-2-furyl benzenemethanol, 3-2-furanyl phenylmethanol, 3-2-furanyl-phenylmethanol, 3-2-furanyl phenyl methanol, 3-2-furyl phenyl methan-1-ol
PubChem CID 11116545
IUPAC Name [3-(furan-2-yl)phenyl]methanol
SMILES C1=CC(=CC(=C1)CO)C2=CC=CO2
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10687854
SDS View Documents
Thermo Scientific Maybridge
CC46109DA
1 g
£95.50
1g
Please to purchase this item. Need a web account? Register with us today!
10624005
SDS View Documents
Thermo Scientific Maybridge
CC46109DE
5 g
£289.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

89929-93-1
174.199
SBTRFADZILHXNG-UHFFFAOYSA-N
11116545
C1=CC(=CC(=C1)CO)C2=CC=CO2
C11H10O2
MFCD06203100
3-2-furyl phenyl methanol, 3-furan-2-yl phenyl methanol, 3-2-furyl benzyl alcohol, 3-2-furyl benzenemethanol, 3-2-furanyl phenylmethanol, 3-2-furanyl-phenylmethanol, 3-2-furanyl phenyl methanol, 3-2-furyl phenyl methan-1-ol
[3-(furan-2-yl)phenyl]methanol

Specifications

89929-93-1
100.0
Amber Glass Bottle
MFCD06203100
SBTRFADZILHXNG-UHFFFAOYSA-N
[3-(furan-2-yl)phenyl]methanol
11116545
95%
94.5
95%
C11H10O2
3-2-furyl phenyl methanol, 3-furan-2-yl phenyl methanol, 3-2-furyl benzyl alcohol, 3-2-furyl benzenemethanol, 3-2-furanyl phenylmethanol, 3-2-furanyl-phenylmethanol, 3-2-furanyl phenyl methanol, 3-2-furyl phenyl methan-1-ol
C1=CC(=CC(=C1)CO)C2=CC=CO2
174.199
174.2
[3-(2-Furyl)phenyl]methanol
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.