missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Aminoethylamino)propyltrimethoxysilane, tech. 90%, Thermo Scientific Chemicals

£22.70 - £190.00

Chemical Identifiers

CAS 1760-24-3
Molecular Formula C8H22N2O3Si
Molecular Weight (g/mol) 222.36
MDL Number MFCD00008173
InChI Key PHQOGHDTIVQXHL-UHFFFAOYSA-N
Synonym n-3-trimethoxysilyl propyl ethylenediamine, n-2-aminoethyl-3-aminopropyltrimethoxysilane, 3-2-aminoethylamino propyltrimethoxysilane, en-aptas, silicone a-1120, prosil 3128, aas-m, n1-3-trimethoxysilyl propyl ethane-1,2-diamine, unii-28zcs5ga8g, 3-2-aminoethyl aminopropyl trimethoxysilane
PubChem CID 15659
IUPAC Name N'-(3-trimethoxysilylpropyl)ethane-1,2-diamine
SMILES CO[Si](CCCNCCN)(OC)OC
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11478816
View Documents
Thermo Scientific Alfa Aesar
A16981.18
50 g
£22.70
50g
Please to purchase this item. Need a web account? Register with us today!
11488816
View Documents
Thermo Scientific Alfa Aesar
A16981.30
250 g
£61.41
250g
Please to purchase this item. Need a web account? Register with us today!
11468816
View Documents
Thermo Scientific Alfa Aesar
A16981.0B
1000 g
£190.00
1000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1760-24-3
222.36
PHQOGHDTIVQXHL-UHFFFAOYSA-N
15659
CO[Si](CCCNCCN)(OC)OC
C8H22N2O3Si
MFCD00008173
n-3-trimethoxysilyl propyl ethylenediamine, n-2-aminoethyl-3-aminopropyltrimethoxysilane, 3-2-aminoethylamino propyltrimethoxysilane, en-aptas, silicone a-1120, prosil 3128, aas-m, n1-3-trimethoxysilyl propyl ethane-1,2-diamine, unii-28zcs5ga8g, 3-2-aminoethyl aminopropyl trimethoxysilane
N'-(3-trimethoxysilylpropyl)ethane-1,2-diamine

Specifications

261°C to 263°C
C8H22N2O3Si
UN2735
136°C (276°F)
PHQOGHDTIVQXHL-UHFFFAOYSA-N
N'-(3-trimethoxysilylpropyl)ethane-1,2-diamine
15659
90%
1.442
1.01
1760-24-3
MFCD00008173
636230
n-3-trimethoxysilyl propyl ethylenediamine, n-2-aminoethyl-3-aminopropyltrimethoxysilane, 3-2-aminoethylamino propyltrimethoxysilane, en-aptas, silicone a-1120, prosil 3128, aas-m, n1-3-trimethoxysilyl propyl ethane-1,2-diamine, unii-28zcs5ga8g, 3-2-aminoethyl aminopropyl trimethoxysilane
CO[Si](CCCNCCN)(OC)OC
222.36
222.36
Technical
Air and moisture sensitive
3-(2-Aminoethylamino)propyltrimethoxysilane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P261-P264b-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P362-P501c

H315-H317-H318

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

missing translation for 'einecsNumber' : 217-164-6

missing translation for 'rtecsNumber' : KV7400000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only