missing translation for 'onlineSavingsMsg'
Learn More

2-(Trimethylsilyl)thiazole, 97%, Thermo Scientific Chemicals

Product Code. 11434977
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79265-30-8
Molecular Formula C6H11NSSi
Molecular Weight (g/mol) 157.31
MDL Number MFCD00066274
InChI Key VJCHUDDPWPQOLH-UHFFFAOYSA-N
Synonym 2-trimethylsilyl thiazole, 2-trimethylsilylthiazole, 2-trimethylsilyl-1,3-thiazole, thiazole, 2-trimethylsilyl, 2-trimethylsilanylthiazole, trimethyl 1,3-thiazol-2-yl silane, 2-tms-thiazole, pubchem10262, acmc-209pgq
PubChem CID 588453
IUPAC Name trimethyl(1,3-thiazol-2-yl)silane
SMILES C[Si](C)(C)C1=NC=CS1

Specifications

Density 0.985
Boiling Point 53°C to 55°C (10 mmHg)
Flash Point 57°C (135°F)
Refractive Index 1.497
Quantity 1 g
UN Number UN1993
Sensitivity Moisture sensitive
Formula Weight 157.31
Percent Purity 97%
Chemical Name or Material 2-(Trimethylsilyl)thiazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'