missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-pyrrolidin-1-ylaniline, Thermo Scientific™
£23.00 - £155.00
Chemical Identifiers
CAS | 21627-58-7 |
---|---|
Molecular Formula | C10H14N2 |
Molecular Weight (g/mol) | 162.236 |
InChI Key | BFPUBGCFJMIZDF-UHFFFAOYSA-N |
Synonym | 2-pyrrolidin-1-yl aniline, 2-pyrrolidinoaniline, 2-pyrrolidin-1-yl-phenylamine, 2-pyrrolidinylphenylamine, 2-pyrrolizinoaniline, acmc-209fmh, 2-pyrrolidin-1-ylphenylamine, 1-2-aminophenyl pyrrolidine, 2-pyrrolidin-1-ylphenyl amine, 2-1-pyrrolidinyl-benzenamine |
PubChem CID | 937501 |
IUPAC Name | 2-pyrrolidin-1-ylaniline |
SMILES | C1CCN(C1)C2=CC=CC=C2N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10295784
|
Thermo Scientific Maybridge
CC44814CB |
250 mg |
£23.00
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10091284
|
Thermo Scientific Maybridge
CC44814DA |
1 g |
£54.10
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10777475
|
Thermo Scientific Maybridge
CC44814DE |
5 g |
£155.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
21627-58-7 | |
162.236 | |
2-pyrrolidin-1-yl aniline, 2-pyrrolidinoaniline, 2-pyrrolidin-1-yl-phenylamine, 2-pyrrolidinylphenylamine, 2-pyrrolizinoaniline, acmc-209fmh, 2-pyrrolidin-1-ylphenylamine, 1-2-aminophenyl pyrrolidine, 2-pyrrolidin-1-ylphenyl amine, 2-1-pyrrolidinyl-benzenamine | |
2-pyrrolidin-1-ylaniline |
C10H14N2 | |
BFPUBGCFJMIZDF-UHFFFAOYSA-N | |
937501 | |
C1CCN(C1)C2=CC=CC=C2N |
Specifications
21627-58-7 | |
C10H14N2 | |
BFPUBGCFJMIZDF-UHFFFAOYSA-N | |
2-pyrrolidin-1-ylaniline | |
937501 | |
97% |
97% | |
2-pyrrolidin-1-yl aniline, 2-pyrrolidinoaniline, 2-pyrrolidin-1-yl-phenylamine, 2-pyrrolidinylphenylamine, 2-pyrrolizinoaniline, acmc-209fmh, 2-pyrrolidin-1-ylphenylamine, 1-2-aminophenyl pyrrolidine, 2-pyrrolidin-1-ylphenyl amine, 2-1-pyrrolidinyl-benzenamine | |
C1CCN(C1)C2=CC=CC=C2N | |
162.236 | |
162.23 | |
2-Pyrrolidin-1-ylaniline |
Safety and Handling
GHS Signal Word: Warning
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title