missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Propanol, puriss. p.a., Reag. ACS, Reag. ISO, Reag. PH. EUR., Reag. USP, Honeywell Riedel-de Haën™
puriss. p.a., Reag. ACS, Reag. ISO, Reag. PH. EUR., Reag. USP
£41.27 - £98.73
Chemical Identifiers
CAS | 67-63-0 |
---|---|
Molecular Formula | C3H8O |
Molecular Weight (g/mol) | 60.096 |
InChI Key | KFZMGEQAYNKOFK-UHFFFAOYSA-N |
Synonym | isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol |
PubChem CID | 3776 |
ChEBI | CHEBI:17824 |
IUPAC Name | propan-2-ol |
SMILES | CC(C)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15676890
|
Honeywell-Riedel-de Haen
72299-1L |
1 L | Plastic Bottle |
£41.27
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15686890
|
Honeywell-Riedel-de Haen
72299-2.5L |
2.5 L | Poly Bottle | N/A | |||||
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Chemical Identifiers
67-63-0 | |
60.096 | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
CHEBI:17824 | |
CC(C)O |
C3H8O | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
3776 | |
propan-2-ol |
Specifications
67-63-0 | |
0.784 - 0.786 g/cm3 | |
12°C | |
UN1219 | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
CC(C)O | |
60.096 | |
CHEBI:17824 | |
>99.9% | |
2-Propanol, puriss. p.a., Reag. ACS, Reag. ISO, Reag. PH. EUR., Reag. USP |
100.0000% | |
77°C to 83°C | |
C3H8O | |
635639 | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
propan-2-ol | |
3776 | |
60.11 | |
ACS Reagent |
Safety and Handling
missing translation for 'shelfLife' : 1080 days from date of manufacture
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Isopropanol
Recommended Storage : Room Temp
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 2-Propanol
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title