missing translation for 'onlineSavingsMsg'
Learn More

2-Picolyl chloride hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number p-184672
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
6959-47-3
C6H6ClN·HCl
164.04
MFCD00012811
JPMRGPPMXHGKRO-UHFFFAOYSA-N
2-chloromethyl pyridine hydrochloride, 2-picolyl chloride hydrochloride, 2-chloromethylpyridine hydrochloride, picolyl chloride hydrochloride, 2-chloromethyl pyridine hcl, 2-pyridylmethylchloride hydrochloride, 2-chloromethyl pyridinium chloride, pyridine, 2-chloromethyl-, hydrochloride, unii-yni4417kx0
23392
CHEBI:76600
2-(chloromethyl)pyridine;hydrochloride
C1=CC=NC(=C1)CCl.Cl
This item is not returnable. View return policy
6959-47-3
C6H6ClN·HCl
164.04
MFCD00012811
JPMRGPPMXHGKRO-UHFFFAOYSA-N
2-chloromethyl pyridine hydrochloride, 2-picolyl chloride hydrochloride, 2-chloromethylpyridine hydrochloride, picolyl chloride hydrochloride, 2-chloromethyl pyridine hcl, 2-pyridylmethylchloride hydrochloride, 2-chloromethyl pyridinium chloride, pyridine, 2-chloromethyl-, hydrochloride, unii-yni4417kx0
23392
CHEBI:76600
2-(chloromethyl)pyridine;hydrochloride
C1=CC=NC(=C1)CCl.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 119.0°C to 125.0°C
Color Brown to Green-Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Formula Weight 164.04
Percent Purity 98%
Physical Form Crystalline Mass
Chemical Name or Material 2-Picolyl chloride hydrochloride, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Picolyl chloride hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'