Learn More
2-Methylcyclohexanol, 99%, mixture of cis and trans, Thermo Scientific Chemicals
£27.03 - £150.75
Chemical Identifiers
CAS | 583-59-5 |
---|---|
Molecular Formula | C7H14O |
Molecular Weight (g/mol) | 114.19 |
MDL Number | MFCD00001443 |
InChI Key | NDVWOBYBJYUSMF-UHFFFAOYSA-N |
Synonym | 2-methylcyclohexanol, cyclohexanol, 2-methyl, o-methylcyclohexanol, 1-methyl-2-cyclohexanol, 2-methyl-1-cyclohexanol, 2-methylcyclohexyl alcohol, cyclohexanol, o-methyl, cis-2-methylcyclohexanol, 2-methyl-cyclohexanol, 2-methylcyclohexanol, mixed isomers |
PubChem CID | 11418 |
IUPAC Name | 2-methylcyclohexan-1-ol |
SMILES | CC1CCCCC1O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10522191
|
Thermo Scientific Acros
166892500 |
250 g |
£27.03
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10479080
|
Thermo Scientific Acros
166890010 |
1 kg |
£70.15
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10449573
|
Thermo Scientific Acros
166890025 |
2.5 kg |
£150.75
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
583-59-5 | |
114.19 | |
NDVWOBYBJYUSMF-UHFFFAOYSA-N | |
11418 | |
CC1CCCCC1O |
C7H14O | |
MFCD00001443 | |
2-methylcyclohexanol, cyclohexanol, 2-methyl, o-methylcyclohexanol, 1-methyl-2-cyclohexanol, 2-methyl-1-cyclohexanol, 2-methylcyclohexyl alcohol, cyclohexanol, o-methyl, cis-2-methylcyclohexanol, 2-methyl-cyclohexanol, 2-methylcyclohexanol, mixed isomers | |
2-methylcyclohexan-1-ol |
Specifications
583-59-5 | |
100.0 | |
Colorless | |
163.0°C to 166.0°C | |
Authentic | |
Glass bottle | |
CH3C6H10OH | |
MFCD00001443 | |
08,303 | |
2-methylcyclohexanol, cyclohexanol, 2-methyl, o-methylcyclohexanol, 1-methyl-2-cyclohexanol, 2-methyl-1-cyclohexanol, 2-methylcyclohexyl alcohol, cyclohexanol, o-methyl, cis-2-methylcyclohexanol, 2-methyl-cyclohexanol, 2-methylcyclohexanol, mixed isomers | |
NDVWOBYBJYUSMF-UHFFFAOYSA-N | |
2-methylcyclohexan-1-ol | |
11418 | |
99% | |
2-Methylcyclohexanol |
98.5 | |
-38.0°C | |
0.9300g/mL | |
58°C | |
98.5% min. (GC) | |
C7H14O | |
1.4615 to 1.4635 | |
06, 11 | |
0.93 | |
Solubility in water: slightly soluble | |
CC1CCCCC1O | |
114.19 | |
114.19 | |
Liquid |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 209-512-
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.