missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-methyl-1,3-oxazole-4-carbaldehyde, Thermo Scientific™
Chemical Identifiers
CAS | 113732-84-6 |
---|---|
Molecular Formula | C5H5NO2 |
Molecular Weight (g/mol) | 111.1 |
InChI Key | ARAUEWKXKTYCHZ-UHFFFAOYSA-N |
Synonym | 2-methyloxazole-4-carbaldehyde, 2-methyl-oxazole-4-carbaldehyde, 2-methyloxazole-4-carboxaldehyde, 2-methyl-4-formyloxazole, 4-oxazolecarboxaldehyde, 2-methyl, 4-formyl-2-methyl-1,3-oxazole, 2-methyl-1,3-oxazole-4-carboxaldehyde, acmc-20ai0d, 2-methyl-4-oxazolecarbaldehyde |
PubChem CID | 11073372 |
IUPAC Name | 2-methyl-1,3-oxazole-4-carbaldehyde |
SMILES | CC1=NC(=CO1)C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11346975
|
Thermo Scientific Maybridge
CC57904CB |
250 mg | Amber Glass Bottle |
N/A
|
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10515100
|
Thermo Scientific Maybridge
CC57904DA |
1 g | Amber Glass Bottle |
N/A
|
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11356975
|
Thermo Scientific Maybridge
CC57904DE |
5 g | Amber Glass Bottle |
N/A
|
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
113732-84-6 | |
111.1 | |
2-methyloxazole-4-carbaldehyde, 2-methyl-oxazole-4-carbaldehyde, 2-methyloxazole-4-carboxaldehyde, 2-methyl-4-formyloxazole, 4-oxazolecarboxaldehyde, 2-methyl, 4-formyl-2-methyl-1,3-oxazole, 2-methyl-1,3-oxazole-4-carboxaldehyde, acmc-20ai0d, 2-methyl-4-oxazolecarbaldehyde | |
2-methyl-1,3-oxazole-4-carbaldehyde |
C5H5NO2 | |
ARAUEWKXKTYCHZ-UHFFFAOYSA-N | |
11073372 | |
CC1=NC(=CO1)C=O |
Specifications
113732-84-6 | |
2-methyloxazole-4-carbaldehyde, 2-methyl-oxazole-4-carbaldehyde, 2-methyloxazole-4-carboxaldehyde, 2-methyl-4-formyloxazole, 4-oxazolecarboxaldehyde, 2-methyl, 4-formyl-2-methyl-1,3-oxazole, 2-methyl-1,3-oxazole-4-carboxaldehyde, acmc-20ai0d, 2-methyl-4-oxazolecarbaldehyde | |
CC1=NC(=CO1)C=O | |
111.1 | |
111.1 | |
2-methyl-1, 3-oxazole-4-carbaldehyde |
C5H5NO2 | |
ARAUEWKXKTYCHZ-UHFFFAOYSA-N | |
2-methyl-1,3-oxazole-4-carbaldehyde | |
11073372 | |
97% |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title