Learn More
2-Mercaptoethanol, Electrophoresis Grade, 98+%, Thermo Scientific Chemicals
£27.14 - £49.04
Chemical Identifiers
CAS | 60-24-2 |
---|---|
Molecular Formula | C2H6OS |
Molecular Weight (g/mol) | 78.13 |
MDL Number | MFCD00004890 |
InChI Key | DGVVWUTYPXICAM-UHFFFAOYSA-N |
Synonym | 2-mercaptoethanol, mercaptoethanol, thioglycol, beta-mercaptoethanol, ethanol, 2-mercapto, 2-thioethanol, 2-hydroxy-1-ethanethiol, thioethylene glycol, 2-hydroxyethanethiol, 2-hydroxyethyl mercaptan |
PubChem CID | 1567 |
ChEBI | CHEBI:41218 |
SMILES | OCCS |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15418219
|
Thermo Scientific Alfa Aesar
J66742.30 |
250 g |
£27.14
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15400647
|
Thermo Scientific Alfa Aesar
J66742.0B |
1000 g |
£49.04
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
β-Mercaptoethanol can act as an enzyme reactivator in systems necessitating reduction for activation, and has been commonly used to reduce disulfide bonds in order to separate protein subunits for use in electrophoresis. This product can also be used to form o-Phthalaldehyde-beta-mercaptoethanol (OPAME), a fluorogenic reagent shown to detect amino acids based on its reaction with primary amines. More recently OPAME has been used to distinguish between ssDNA/dsDNA and even single base mismatches in the hybridization of DNA. It is widely used for retarding oxidation of biological compounds in solution.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsβ-Mercaptoethanol can act as an enzyme reactivator in systems necessitating reduction for activation, and has been commonly used to reduce disulfide bonds in order to separate protein subunits for use in electrophoresis. This product can also be used to form o-Phthalaldehyde-beta-mercaptoethanol (OPAME), a fluorogenic reagent shown to detect amino acids based on its reaction with primary amines. More recently OPAME has been used to distinguish between ssDNA/dsDNA and even single base mismatches in the hybridization of DNA. It is widely used for retarding oxidation of biological compounds in solution.
Solubility
Fully miscible in water. Soluble in alcohol, ether, benzene, and other organic solvents.
Notes
Handle under dry protective gas. Store in cool condition. Incompatible with air, moisture, oxidizing agents.
Chemical Identifiers
60-24-2 | |
78.13 | |
DGVVWUTYPXICAM-UHFFFAOYSA-N | |
1567 | |
OCCS |
C2H6OS | |
MFCD00004890 | |
2-mercaptoethanol, mercaptoethanol, thioglycol, beta-mercaptoethanol, ethanol, 2-mercapto, 2-thioethanol, 2-hydroxy-1-ethanethiol, thioethylene glycol, 2-hydroxyethanethiol, 2-hydroxyethyl mercaptan | |
CHEBI:41218 |
Specifications
60-24-2 | |
Colorless | |
156°C to 158°C | |
Unpleasant | |
HSCH2CH2OH | |
UN2966 | |
Air sensitive; Hygroscopic | |
2-mercaptoethanol, mercaptoethanol, thioglycol, beta-mercaptoethanol, ethanol, 2-mercapto, 2-thioethanol, 2-hydroxy-1-ethanethiol, thioethylene glycol, 2-hydroxyethanethiol, 2-hydroxyethyl mercaptan | |
DGVVWUTYPXICAM-UHFFFAOYSA-N | |
2-sulfanylethanol | |
1567 | |
78.13 | |
Electrophoresis | |
2-Mercaptoethanol |
-100°C | |
1.114 g/mL | |
73°C (163°F) | |
C2H6OS | |
MFCD00004890 | |
773648 | |
14,5869 | |
Fully miscible in water. Soluble in alcohol,ether,benzene,and other organic solvents. | |
OCCS | |
78.13 | |
CHEBI:41218 | |
≥98% | |
Liquid |
Safety and Handling
GHS H Statement
H301-H310-H318-H227-H315-H317-H335
Toxic if swallowed.
Fatal in contact with skin.
Causes serious eye damage.
Combustible liquid.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P210-P235-P260-P262-P264b-P270-P271-P272-P280-P301+P310-P302+P350-P304+P340-P305+P351+P338-P310-P311-P314-P330-P333+P313-P361-P363-P370+P378q-P501c
H227-H301+H331-H310-H315-H317-H318-H373
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packaging Group: II; Proper Shipping Name: THIOGLYCOL
missing translation for 'einecsNumber' : 200-464-6
missing translation for 'rtecsNumber' : KL5600000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H301-Toxic if swallowed.
- H310-Fatal in contact with skin.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H361f-Suspected of damaging fertility.
- H373-May cause damage to organs.
- H400-Very toxic to aquatic life.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only