missing translation for 'onlineSavingsMsg'
Learn More

2-Hexanone, Reagent grade, 98%, Honeywell™

Product Code. 15640970
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade, 98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-78-6
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00009482
InChI Key QQZOPKMRPOGIEB-UHFFFAOYSA-N
Synonym 2-hexanone, hexanone, 2-oxohexane, propylacetone, n-butyl methyl ketone, butyl methyl ketone, methyl butyl ketone, methyl n-butyl ketone, hexanone-2, ketone, butyl methyl
PubChem CID 11583
IUPAC Name hexan-2-one
SMILES CCCCC(C)=O

Specifications

Melting Point -57°C
Color Colorless
Density 0.812g/mL at 25°C
Boiling Point 127°C
Flash Point 23°C (73.4°F)
Packaging Glass Bottle
Linear Formula CH3(CH2)3COCH3
Refractive Index n20/D 1.401(lit.)
Quantity 250 g
UN Number UN1224
Beilstein 1737676
Formula Weight 100.16g/mol
Percent Purity 98%
Grade Reagent
Physical Form Liquid
Chemical Name or Material 2-Hexanone
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'