missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorostyrene, 98%, stab. with 0.1% 4-tert-butylcatechol, Thermo Scientific Chemicals

Product Code. 11478007
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It finds its application in the homo- and cross-olefin metathesis coupling of vinylphosphane oxides and electron-poor alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the homo- and cross-olefin metathesis coupling of vinylphosphane oxides and electron-poor alkenes.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents, heat, light, UV radiation and free radical initiators. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 394-46-7
Molecular Formula C8H7F
Molecular Weight (g/mol) 122.142
MDL Number MFCD00013550
InChI Key YNQXOOPPJWSXMW-UHFFFAOYSA-N
Synonym 2-fluorostyrene, o-fluorostyrene, 1-fluoro-2-vinylbenzene, benzene, 1-ethenyl-2-fluoro, benzene, ethenylfluoro, 1-ethenyl-2-fluoro-benzene, 2-fluoro-styrene, acmc-1cuju, 1-fluoro-2-vinyl-benzene
PubChem CID 123056
IUPAC Name 1-ethenyl-2-fluorobenzene
SMILES C=CC1=CC=CC=C1F

Specifications

Density 1.02
Boiling Point 29°C to 30°C (4 mmHg)
Flash Point 35°C (95°F)
Odor Recognizable
Refractive Index 1.52
Quantity 5 g
UN Number UN1993
Beilstein 1925226
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 122.14
Percent Purity 98%
Chemical Name or Material 2-Fluorostyrene, stabilized with 0.1% 4-tert-butylcatechol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.