Learn More
2-Ethyl-1-butanol, 99%, Thermo Scientific Chemicals
£56.50 - £962.00
Chemical Identifiers
CAS | 97-95-0 |
---|---|
Molecular Formula | C6H14O |
Molecular Weight (g/mol) | 102.18 |
MDL Number | MFCD00004744 |
InChI Key | TZYRSLHNPKPEFV-UHFFFAOYSA-N |
Synonym | 2-ethyl-1-butanol, 1-butanol, 2-ethyl, 2-ethylbutanol, 2-ethylbutyl alcohol, pseudohexyl alcohol, 3-methylolpentane, 2-ethylbutanol-1, ethylbutanol, 3-pentylcarbinol, sec-pentylcarbinol |
PubChem CID | 7358 |
IUPAC Name | 2-ethylbutan-1-ol |
SMILES | CCC(CC)CO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11460107
|
Thermo Scientific Alfa Aesar
B21026.AE |
100 mL |
£56.50
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11470107
|
Thermo Scientific Alfa Aesar
B21026.AP |
500 mL |
£242.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11480107
|
Thermo Scientific Alfa Aesar
B21026.0F |
2500 mL |
£962.00
2500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as a perfuming agent in cosmetics industry. 2-Ethyl-1-butanol is used for the synthesis of various pharmaceutical compounds, such as novel branched Alkyl carbamates, acting as anticonvulsant agent. It is also a solvent used for various organic synthesis, separation processes and ionic liquids, and solvent mixtures for pharmaceutical applications . It is also used to make penetrating oils, corrosion inhibitors, plasticizers, and perfumes; as cleaning agent for printed circuits; and to improve flow of paints and varnishes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a perfuming agent in cosmetics industry. 2-Ethyl-1-butanol is used for the synthesis of various pharmaceutical compounds, such as novel branched Alkyl carbamates, acting as anticonvulsant agent. It is also a solvent used for various organic synthesis, separation processes and ionic liquids, and solvent mixtures for pharmaceutical applications . It is also used to make penetrating oils, corrosion inhibitors, plasticizers, and perfumes; as cleaning agent for printed circuits; and to improve flow of paints and varnishes.
Solubility
Soluble in water (4 mg/ml at 25°C), alcohol, ether, and most organic solvents.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
97-95-0 | |
102.18 | |
TZYRSLHNPKPEFV-UHFFFAOYSA-N | |
7358 | |
CCC(CC)CO |
C6H14O | |
MFCD00004744 | |
2-ethyl-1-butanol, 1-butanol, 2-ethyl, 2-ethylbutanol, 2-ethylbutyl alcohol, pseudohexyl alcohol, 3-methylolpentane, 2-ethylbutanol-1, ethylbutanol, 3-pentylcarbinol, sec-pentylcarbinol | |
2-ethylbutan-1-ol |
Specifications
97-95-0 | |
146°C to 148°C | |
C6H14O | |
UN1987 | |
2-ethyl-1-butanol, 1-butanol, 2-ethyl, 2-ethylbutanol, 2-ethylbutyl alcohol, pseudohexyl alcohol, 3-methylolpentane, 2-ethylbutanol-1, ethylbutanol, 3-pentylcarbinol, sec-pentylcarbinol | |
CCC(CC)CO | |
102.18 | |
102.18 | |
2-Ethyl-1-butanol |
-52°C | |
58°C (136°F) | |
MFCD00004744 | |
1731254 | |
TZYRSLHNPKPEFV-UHFFFAOYSA-N | |
2-ethylbutan-1-ol | |
7358 | |
99% |
Safety and Handling
GHS H Statement
H311-H226-H302
Toxic in contact with skin.
Flammable liquid and vapour.
Harmful if swallowed.
GHS P Statement
P210-P280-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
missing translation for 'rtecsNumber' : EL3850000
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.