missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethoxyethanol, ≥99%, Spectrophotometric Grade, Honeywell™ Riedel-de-Haën™
£85.41 - £147.09
Chemical Identifiers
CAS | 110-80-5 |
---|---|
Molecular Formula | C4H10O2 |
Molecular Weight (g/mol) | 90.12 |
MDL Number | MFCD00002869 |
InChI Key | ZNQVEEAIQZEUHB-UHFFFAOYSA-N |
Synonym | cellosolve, oxitol, ethyl cellosolve, ethylene glycol monoethyl ether, ethanol, 2-ethoxy, ethyl glycol, emkanol, hydroxy ether, ethylene glycol ethyl ether, dowanol ee |
PubChem CID | 8076 |
ChEBI | CHEBI:46788 |
IUPAC Name | 2-ethoxyethan-1-ol |
SMILES | CCOCCO |
Description
Chemical Identifiers
110-80-5 | |
90.12 | |
ZNQVEEAIQZEUHB-UHFFFAOYSA-N | |
8076 | |
2-ethoxyethan-1-ol |
C4H10O2 | |
MFCD00002869 | |
cellosolve, oxitol, ethyl cellosolve, ethylene glycol monoethyl ether, ethanol, 2-ethoxy, ethyl glycol, emkanol, hydroxy ether, ethylene glycol ethyl ether, dowanol ee | |
CHEBI:46788 | |
CCOCCO |
Specifications
110-80-5 | |
0.14 | |
460°F | |
C2H5OCH2CH2OH | |
UN1171 | |
cellosolve, oxitol, ethyl cellosolve, ethylene glycol monoethyl ether, ethanol, 2-ethoxy, ethyl glycol, emkanol, hydroxy ether, ethylene glycol ethyl ether, dowanol ee | |
CCOCCO | |
90.12 | |
3.1 (vs air) | |
90.12g/mol | |
≥99% | |
2-Ethoxyethanol |
0.93g/mL at 25°C | |
42°C (107.6°F) | |
C4H10O2 | |
MFCD00002869 | |
1098271 | |
ZNQVEEAIQZEUHB-UHFFFAOYSA-N | |
2-ethoxyethan-1-ol | |
8076 | |
CHEBI:46788 | |
3.8mmHg (20°C) | |
Sprectrophotometric |
Safety and Handling
P201-P261-P311
H226-H302-H331-H360
missing translation for 'dotInformation' : Class 3 - PG 3 - Ethylene glycol monoethyl ether
missing translation for 'einecsNumber' : 203-804-1
missing translation for 'rtecsNumber' : KK8050000
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title