missing translation for 'onlineSavingsMsg'
Learn More

2-(Dimethylamino)-3-quinolinecarbaldehyde, 97+%, Thermo Scientific™

£19.60 - £156.00

Chemical Identifiers

CAS 728035-61-8
Molecular Formula C12H12N2O
Molecular Weight (g/mol) 200.24
MDL Number MFCD06010203
InChI Key RCRLBHNGTPRGJQ-UHFFFAOYSA-N
PubChem CID 1076116
IUPAC Name 2-(dimethylamino)quinoline-3-carbaldehyde
SMILES CN(C)C1=C(C=O)C=C2C=CC=CC2=N1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10299212
SDS View Documents
Thermo Scientific Maybridge
MO07114CB
250 mg
£19.60
250mg
Please to purchase this item. Need a web account? Register with us today!
10209222
SDS View Documents
Thermo Scientific Maybridge
MO07114DA
1 g
£51.80
1g
Please to purchase this item. Need a web account? Register with us today!
10507474
SDS View Documents
Thermo Scientific Maybridge
MO07114DE
5 g
£156.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

728035-61-8
200.24
RCRLBHNGTPRGJQ-UHFFFAOYSA-N
2-(dimethylamino)quinoline-3-carbaldehyde
C12H12N2O
MFCD06010203
1076116
CN(C)C1=C(C=O)C=C2C=CC=CC2=N1

Specifications

728035-61-8
C12H12N2O
2-dimethylamino quinoline-3-carbaldehyde, 2-dimethylamino-quinoline-3-carbaldehyde, 2-dimethylamino-3-quinolinecarbaldehyde, 3-quinolinecarboxaldehyde,2-dimethylamino, 2-dimethylamino-3-quinoline carbaldehyde, 2-dimethylamino quinoline-3-carboxaldehyde, 3-quinolinecarboxaldehyde, 2-dimethylamino
CN(C)C1=C(C=O)C=C2C=CC=CC2=N1
200.24
200.24
2-(Dimethylamino)-3-quinolinecarbaldehyde
97%
MFCD06010203
RCRLBHNGTPRGJQ-UHFFFAOYSA-N
2-(dimethylamino)quinoline-3-carbaldehyde
1076116
97%
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.