missing translation for 'onlineSavingsMsg'
Learn More

2-Dicyanomethylene-1,3-indanedione, 98%, Thermo Scientific™

Product Code. 15478889
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 16954-74-8
Melting Point ∽270°C (decomposition)
Molecular Formula C12H4N2O2
MDL Number MFCD00051930
Quantity 5 g
UN Number UN3439
Beilstein 2213594
Synonym 2-dicyanomethylene indan-1,3-dione, 2-1,3-dioxo-1h-inden-2 3h-ylidene malononitrile, 2-1,3-dioxoinden-2-ylidene propanedinitrile, acmc-209r6k, 2-dicyanomethyleneindane-1,3-dione, propanedinitrile, 1,3-dihydro-1,3-dioxo-2h-inden-2-ylidene, 2-dicyanomethylene-1,3-indandione, 2-dicyanomethylene indene-1,3-dione, d2,a-indanmalononitrile, 1,3-dioxo, 2-1,3-dioxo-inden-2-ylidene-propanedinitrile
InChI Key WQFLMIPTYDGFOB-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C(=O)C(=C(C#N)C#N)C2=O
IUPAC Name 2-(1,3-dioxoinden-2-ylidene)propanedinitrile
Molecular Weight (g/mol) 208.176
PubChem CID 542782
Formula Weight 208.18
Percent Purity 98%
Chemical Name or Material 2-Dicyanomethylene-1,3-indanedione
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.